Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoechst™ 2495, 90%, Thermo Scientific™

Catalog No. AC230471000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC230471000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC230471000 Supplier Thermo Scientific Chemicals Supplier No. 230471000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72845-94-4
Molecular Formula C20H13NO10S2·2C2H7NO
Synonym Benzoxanthene Yellow H 2495 Fluorochrome

Specifications

Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 90%
Quantity 100 mg
Formula Weight 613.62
Physical Form Crystalline Powder
Chemical Name or Material Hoechst™ 2495, 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.