accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOSNFFFHClHCl

CAS RN 754240-09-0

IUPAC Name: 2-[(morpholin-4-yl)methyl]-5-[(5-{[7-(trifluoromethyl)quinolin-4-yl]sulfanyl}pentyl)oxy]-4H-pyran-4-one dihydrochloride
Synonyms: 2-(morpholin-4-ylmethyl)-5-[(5-{[7-(trifluoromethyl)quinolin-4-yl]sulfanyl}pentyl)oxy]pyran-4-one dihydrochloride
Molecular Weight (g/mol): 581.47
Molecular Formula: C25H29Cl2F3N2O4S
InChi Key: LSECOAJFCKFQJG-UHFFFAOYSA-N
SMILES: Cl.Cl.FC(F)(F)C1=CC=C2C(SCCCCCOC3=COC(CN4CCOCC4)=CC3=O)=CC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)