accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76-62-0

IUPAC Name: 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Synonyms: 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-2-benzofuran-1-one
Molecular Weight (g/mol): 633.91
Molecular Formula: C20H10Br4O4
InChi Key: OBRGVMYQZVQHGO-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(C=C1Br)C1(OC(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results