accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 762-75-4

CH3CH3CH3OO

CAS RN 762-75-4

IUPAC Name: tert-butyl formate
Synonyms: tert-butyloxycarbonyl group
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: RUPAXCPQAAOIPB-UHFFFAOYSA-N
SMILES: CC(C)(C)OC=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)