accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 781652-57-1

IUPAC Name: 3-cyano-N-(1,3-diphenyl-1H-pyrazol-5-yl)benzamide
Synonyms: cdppb
Molecular Weight (g/mol): 364.41
Molecular Formula: C23H16N4O
InChi Key: BKUIZWILNWHFHD-UHFFFAOYSA-N
SMILES: O=C(NC1=CC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results