CAS RN 827022-33-3
IUPAC Name:
2-hydroxyethane-1-sulfonic acid; 6-acetyl-8-cyclopentyl-5-methyl-2-{[5-(piperazin-1-yl)pyridin-2-yl]amino}-7H,8H-pyrido[2,3-d]pyrimidin-7-one
Synonyms:
isethionic acid; palbociclib
Molecular Weight (g/mol):
573.67
Molecular Formula:
C26H35N7O6S
InChi Key:
LYYVFHRFIJKPOV-UHFFFAOYSA-N
SMILES:
OCCS(O)(=O)=O.CC(=O)C1=C(C)C2=CN=C(NC3=NC=C(C=C3)N3CCNCC3)N=C2N(C2CCCC2)C1=O
1
–
1
of
1
results
1
R&D Systems™ PD 0332991 isethionate
CAS: 827022-33-3 Molecular Formula: C26H35N7O6S