accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOClOHONa

CAS RN 828934-41-4

IUPAC Name: 2-[6-(4-chlorophenoxy)hexyl]oxirane-2-carboxylic acid sodium
Synonyms: 2-[6-(4-chlorophenoxy)hexyl]oxirane-2-carboxylic acid sodium
Molecular Weight (g/mol): 321.75
Molecular Formula: C15H19ClNaO4
InChi Key: ZUMIFLJZQDMWAU-UHFFFAOYNA-N
SMILES: [Na].OC(=O)C1(CCCCCCOC2=CC=C(Cl)C=C2)CO1
Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)