accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 844499-71-4

IUPAC Name: 4-hydroxy-3-{2'-hydroxy-[1,1'-biphenyl]-4-yl}-6-oxo-6H,7H-thieno[2,3-b]pyridine-5-carbonitrile
Synonyms: 4-hydroxy-3-{2'-hydroxy-[1,1'-biphenyl]-4-yl}-6-oxo-7H-thieno[2,3-b]pyridine-5-carbonitrile
Molecular Weight (g/mol): 360.39
Molecular Formula: C20H12N2O3S
InChi Key: CTESJDQKVOEUOY-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C1=CC=C(C=C1)C1=CSC2=C1C(O)=C(C#N)C(=O)N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results