accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNHOHNSOOFCl

CAS RN 852918-02-6

IUPAC Name: N-{[4-(N'-benzoylhydrazinecarbonyl)phenyl]methyl}-3-chloro-4-fluorobenzene-1-sulfonamide
Synonyms: N-{[4-(N'-benzoylhydrazinecarbonyl)phenyl]methyl}-3-chloro-4-fluorobenzenesulfonamide
Molecular Weight (g/mol): 461.89
Molecular Formula: C21H17ClFN3O4S
InChi Key: FYIBXBFDXNPBSF-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=C(C=C1)S(=O)(=O)NCC1=CC=C(C=C1)C(=O)NNC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results