accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 864756-35-4

IUPAC Name: 4-[(4-fluorophenyl)methoxy]-1-{4-[2-(pyrrolidin-1-yl)ethoxy]phenyl}-1,2-dihydropyridin-2-one
Synonyms: 4-[(4-fluorophenyl)methoxy]-1-{4-[2-(pyrrolidin-1-yl)ethoxy]phenyl}pyridin-2-one
Molecular Weight (g/mol): 408.47
Molecular Formula: C24H25FN2O3
InChi Key: ANCFKYJMXNMYNZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(COC2=CC(=O)N(C=C2)C2=CC=C(OCCN3CCCC3)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results