CAS RN 869185-85-3
IUPAC Name:
4-[4-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-1H-imidazol-5-yl]pyridine hydrochloride
Synonyms:
4-[5-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-3H-imidazol-4-yl]pyridine hydrochloride
Molecular Weight (g/mol):
413.9
Molecular Formula:
C21H17ClFN3OS
InChi Key:
WOSGGXINSLMASH-UHFFFAOYNA-N
SMILES:
Cl.CS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
R&D Systems™ SB 203580 hydrochloride
CAS: 869185-85-3 Molecular Formula: C21H17ClFN3OS