CAS RN 869497-75-6
IUPAC Name:
6-chloro-2-(4-methylpiperazine-1-carbonyl)-1H-1,3-benzodiazole; formic acid; prop-2-enoic acid
Synonyms:
5-chloro-2-(4-methylpiperazine-1-carbonyl)-3H-1,3-benzodiazole; acrylic acid; formic acid
Molecular Weight (g/mol):
396.83
Molecular Formula:
C17H21ClN4O5
InChi Key:
BODOIXYJOORQBQ-UHFFFAOYSA-N
SMILES:
OC=O.OC(=O)C=C.CN1CCN(CC1)C(=O)C1=NC2=CC=C(Cl)C=C2N1
1
–
1
of
1
results
1
R&D Systems™ JNJ 10191584 maleate
CAS: 869497-75-6 Molecular Formula: C17H21ClN4O5