CAS RN 873054-44-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ CFTR Potentiator, VX-770
A cell-permeable, potentiator of CFTR that directly, reversibly, and allosterically binds to WT and G551D mutant forms (≈10 μM) and increases the open probability to restore lung function.
2
Tocris Bioscience™ Ivacaftor
Potent and selective CFTR potentiator