accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONHClONHCl(E/Z)

CAS RN 917837-54-8

IUPAC Name: N-(4-chlorophenyl)-3-[(4-chlorophenyl)amino]-2-(3-methyl-1,2-oxazol-5-yl)prop-2-enamide
Synonyms: N-(4-chlorophenyl)-3-[(4-chlorophenyl)amino]-2-(3-methyl-1,2-oxazol-5-yl)prop-2-enamide
Molecular Weight (g/mol): 388.25
Molecular Formula: C19H15Cl2N3O2
InChi Key: VMAKIACTLSBBIY-UHFFFAOYSA-N
SMILES: CC1=NOC(=C1)C(=CNC1=CC=C(Cl)C=C1)C(=O)NC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results