CAS RN 925434-55-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ SRT1720
SRT1720, CAS 925434-55-5, is a cell-permeable inhibitor of the mitochondrial SIRT3. Inhibition is AceCS2-competitive (Ki = 0.56μM; Km = 2.44μM), but NAD+-uncompetitive (Ki = 0.34μM; Km = 280μM).
2
Tocris Bioscience™ SRT 1720
Selective SIRT1 activator; induces autophagy and mitochondrial biogenesis