accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOONH2

CAS RN 942289-87-4

IUPAC Name: 8-(4-aminophenyl)-2-(morpholin-4-yl)-4H-chromen-4-one
Synonyms: 8-(4-aminophenyl)-2-(morpholin-4-yl)chromen-4-one
Molecular Weight (g/mol): 322.36
Molecular Formula: C19H18N2O3
InChi Key: WUKMIBOGGXMBAC-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=C2OC(=CC(=O)C2=CC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)