CAS RN 99201-87-3
IUPAC Name:
2-chloro-6-(piperidin-4-ylsulfanyl)pyridine hydrochloride
Synonyms:
2-chloro-6-(piperidin-4-ylsulfanyl)pyridine hydrochloride
Molecular Weight (g/mol):
265.2
Molecular Formula:
C10H14Cl2N2S
InChi Key:
GRXDJABVNGUGCW-UHFFFAOYSA-N
SMILES:
Cl.ClC1=CC=CC(SC2CCNCC2)=N1
1
–
1
of
1
results
1
R&D Systems™ Anpirtoline hydrochloride
CAS: 99201-87-3 Molecular Formula: C10H14Cl2N2S