CAS RN 99614-01-4
IUPAC Name:
hydrogen 9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-2,3,4,9-tetrahydro-1H-carbazol-4-one chloride
Synonyms:
hydrogen ondansetron chloride
Molecular Weight (g/mol):
329.83
Molecular Formula:
C18H20ClN3O
InChi Key:
MKBLHFILKIKSQM-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CN1C2=C(C3=CC=CC=C13)C(=O)C(CN1C=CN=C1C)CC2
1
–
1
of
1
results
1
R&D Systems™ Ondansetron hydrochloride
CAS: 99614-01-4 Molecular Formula: C18H20ClN3O