accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 998-00-5

CH3H3CAlCH3H3COAlH3CCH3CH3CH3

CAS RN 998-00-5

IUPAC Name: {[bis(2-methylpropyl)alumanyl]oxy}bis(2-methylpropyl)alumane
Synonyms: {[bis(2-methylpropyl)alumanyl]oxy}bis(2-methylpropyl)alumane
Molecular Weight (g/mol): 298.43
Molecular Formula: C16H36Al2O
InChi Key: CTNFGBKAHAEKFE-UHFFFAOYSA-N
SMILES: CC(C)C[Al](CC(C)C)O[Al](CC(C)C)CC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results