Gloves
Protective gloves and sleeves available in an array of styles and materials for applications such as lab, electrical, medical, and cleanroom work. Protects against hazardous substances, extreme temperatures, impact, sharp objects, and more.
Selection of the appropriate hand protection is based on the hazards workers encounter, such as skin absorption of harmful substances, severe cuts, lacerations, abrasions, punctures, burns, and harmful temperature extremes.
Disposable Gloves
Disposable gloves are typically made from materials such as latex, nitrile, neoprene, butyl, vinyl or specialty formulations that offer some form of chemical resistance. Their thinner construction, like what is found in exam gloves or finger cots, enhances touch sensitivity for tasks involving fine motor skills. They are mostly ambidextrous, but in some applications (i.e., cleanroom gloves) they are pair packed. They are available sterile or non-sterile.
Reusable Gloves
Like disposable gloves, reusable gloves come in a variety of materials and styles designed to handle more demanding hazards. Unlike disposables, reusables can be constructed of one or more materials or fabrics that offer enhanced hand protection.
- Work gloves and mechanics’ gloves may have a soft inner lining and an outer layer of heavier cloth, leather, or synthetic material with reinforced palms or knuckles
- Chemical-resistant gloves may be “dipped” or coated with a layer of nitrile, neoprene, or other materials
- Cut-resistant gloves may be woven of specialty fibers that afford cut or puncture resistance; some styles can be worn underneath other gloves to offer dual protection
- Firefighting, rescue, and tactical gloves may include layers to protect wearers from punctures or extreme temperatures
Other Hand Protection Options
Other reusable glove designs are available for handling more specialized hazards:
- Cold-resistant gloves
- Heat-resistant gloves
- Welding and electrical safety gloves
- Dry box gloves
Filtered Search Results
Ansell Kimtech™ Purple Nitrile™ Gloves
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Purple nitrile glove offers maximum protection for higher-risk procedures. Unique color differentiates from latex and other synthetic gloves.
Tensile Strength | 21 MPa |
---|---|
Disposable | Single-use |
Length (Metric) | 24.13 cm |
Applications | Laboratory,Healthcare |
Chemotherapeutic Rated | Yes |
Finger Thickness (English) | 6 mil |
Color | Purple |
Length (English) | 9.5 in. |
Material | Nitrile |
Powdered | Powder Free |
Finger Thickness (Metric) | 0.15 mm |
Thickness (Metric) | 0.15 mm |
Series | Kimtech™ |
FDA 510 (k) | Yes |
Thickness (English) | 6 mil |
Palm Thickness (English) | 4.7 mil |
Latex Free | Yes |
Exterior Finish | Partially Textured |
Certifications/Compliance | ASTM D6978-05 (non-sterile only), Meets USP <800> guidelines |
AQL | 0.65 |
Palm Thickness (Metric) | 0.12 mm |
Fisherbrand™ Powder Free Nitrile Gloves
Choose a latex-free glove with exceptional durability and puncture resistance. Fisherbrand™ powder-free Nitrile Exam Gloves provide a superior fit, comfort, tactile sensitivity and dexterity.
Disposable | Single-use |
---|---|
Length (Metric) | 245 mm |
Applications | Healthcare,Industrial,Laboratory |
Chemotherapeutic Rated | No |
Finger Thickness (English) | 4.7 mil |
Color | Blue |
Length (English) | 9.6 in. |
Quantity | 100 Gloves/Box |
Material | Nitrile |
Powdered | Powder Free |
Sterility | Non-sterile |
Finger Thickness (Metric) | 0.12 mm |
Thickness (Metric) | Palm: 0.09 mm, Finger: 0.12 mm |
FDA 510 (k) | Yes |
Thickness (English) | Palm: 3.5 mil, Finger: 4.7 mil |
Palm Thickness (English) | 3.5 mil |
Latex Free | Yes |
Exterior Finish | Fully Textured |
Certifications/Compliance | ASTM D6319,FDA21 CFR 177-2600-US Food Contact Approved,US QSR/FDA 510(k) Medical Examination Grade |
AQL | 1.5 |
Palm Thickness (Metric) | 0.09 mm |
Fisherbrand™ Comfort Nitrile Gloves with Aloe and Chamomile
Soft nitrile protection with aloe and chamomile.
Disposable | Single-use |
---|---|
Length (Metric) | 250 mm |
Applications | Healthcare,Industrial,Laboratory |
Chemotherapeutic Rated | Yes |
Finger Thickness (English) | 4.7 mil |
Color | Blue,Green |
Length (English) | 9.8 in. |
Quantity | 100 Gloves/Box |
Material | Nitrile |
Powdered | Powder Free |
Sterility | Non-sterile |
Finger Thickness (Metric) | 0.12 mm |
Thickness (Metric) | Palm: 0.09 mm, Finger: 0.12 mm |
FDA 510 (k) | Yes |
Product Type | Nitrile Examination Glove |
Thickness (English) | Palm: 3.5 mil, Finger: 4.7 mil |
Palm Thickness (English) | 3.5 mil |
Latex Free | Yes |
Exterior Finish | Fingertip Textured |
Certifications/Compliance | ASTM D6319,ASTM D6978,FDA21 CFR 177-2600, US Food Contact Approved,US QSR/FDA 510(k) Medical Examination Grade |
AQL | 1.5 |
Glove Cuff Style | Beaded |
Palm Thickness (Metric) | 0.09 mm |
Fisherbrand™ Comfort Nitrile Gloves
Choose a glove with enhanced comfort and tactile sensitivity. Fisherbrand™ Comfort Nitrile Gloves are powder free and latex-free with a beaded fit and textured fingers.
Disposable | Single-use |
---|---|
Applications | Healthcare,Industrial,Laboratory |
Chemotherapeutic Rated | Yes |
Finger Thickness (English) | 3.5 mil |
Color | Blue |
Length (English) | 9.8 in. |
Material | Nitrile |
Powdered | Powder Free |
Sterility | Non-sterile |
Hand | Ambidextrous |
Finger Thickness (Metric) | 0.09 mm |
Thickness (Metric) | Palm: 0.07 mm, Finger: 0.09 mm |
FDA 510 (k) | Yes |
Thickness (English) | Palm: 2.8 mil, Finger: 3.5 mil |
Palm Thickness (English) | 2.8 mil |
Latex Free | Yes |
Exterior Finish | Fingertip Textured |
Certifications/Compliance | ASTM D6319,ASTM D6978,ISO 10993-10:2002, ISO 9001, ISO 13485,FDA21 CFR 177- 2600-US Food Contact Approved,US QSR/FDA 510(k) Medical Examination Grade |
AQL | 1.5 |
Palm Thickness (Metric) | 0.07 mm |
Fisherbrand™ Nitrile Class 100 Cleanroom Non-Sterile Gloves (ISO 5)
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Class 100 (ISO 5) Cleanroom Gloves, ambidextrous, non-sterile, chemical resistance to a range of common chemicals and substances.
Accelerator Free | Yes |
---|---|
Certifications/Compliance (Europe) | EN420:2003+A1:2009, the European Medical Gloves Standard EN455 (Parts 1, 2 & 3) & EN374-1:2016 |
Color | White |
Latex Free | Yes |
Material | Nitrile |
Certifications/Compliance | ISO 5 |
Sterility | Non-sterile |
Fisherbrand™ Powder Free Black Nitrile Gloves
Choose a latex-free glove with exceptional durability and puncture resistance. Fisherbrand™ powder free Nitrile Exam Gloves provide a superior fit, comfort, tactile sensitivity and dexterity.
Disposable | Single-use |
---|---|
Length (Metric) | 245 mm |
Applications | Healthcare,Industrial,Laboratory |
Chemotherapeutic Rated | No |
Finger Thickness (English) | 5.5 mil |
Color | Black |
Length (English) | 9.6 in. |
Quantity | 100 Gloves/Box |
Material | Nitrile |
Powdered | Powder Free |
Sterility | Non-sterile |
Finger Thickness (Metric) | 0.14 mm |
Thickness (Metric) | Palm: 0.09 mm, Finger: 0.14 mm |
FDA 510 (k) | Yes |
Thickness (English) | Palm: 3.5 mil, Finger: 5.5 mil |
Palm Thickness (English) | 3.5 mil |
Latex Free | Yes |
Exterior Finish | Fingertip Textured |
Certifications/Compliance | ASTM D6319,FDA21 CFR 177-2600-US Food Contact Approved,US QSR/FDA 510(k) Medical Examination Grade |
AQL | 1.5 |
Palm Thickness (Metric) | 0.09 mm |
Fisherbrand™ Powder Free Dark Blue Nitrile Gloves
Choose a latex-free glove with exceptional durability and puncture resistance. Fisherbrand™ powder free Nitrile Exam Gloves provide a superior fit, comfort, tactile sensitivity and dexterity.
Disposable | Single-use |
---|---|
Length (Metric) | 240 mm |
Applications | Healthcare,Industrial,Laboratory |
Chemotherapeutic Rated | No |
Finger Thickness (English) | 5.5 mil |
Color | Dark Blue |
Length (English) | 9.5 in. |
Quantity | 100 Gloves/Box |
Material | Nitrile |
Powdered | Powder Free |
Sterility | Non-sterile |
Finger Thickness (Metric) | 0.14 mm |
Thickness (Metric) | Palm: 0.12 mm, Finger: 0.14 mm |
FDA 510 (k) | No |
Thickness (English) | Palm: 4.7 mil, Finger: 5.5 mil |
Palm Thickness (English) | 4.7 mil |
Latex Free | Yes |
Exterior Finish | Fully Textured |
Certifications/Compliance | ASTM D6319,FDA21 CFR 177-2600-US Food Contact Approved |
AQL | 1.5 |
Palm Thickness (Metric) | 0.12 mm |
Ansell MICROFLEX™ FreeForm™ SE FFS-700 Nitrile Gloves
Powder free, nitrile exam gloves with textured fingertips offer reliable protection and a secure grip.
Tensile Strength | 32 MPa |
---|---|
Disposable | Single-use |
Length (Metric) | 24.13 cm |
Applications | Healthcare,Laboratory,Industrial |
Chemotherapeutic Rated | No |
Finger Thickness (English) | 5.1 mil |
Color | Blue |
Length (English) | 9.5 in. |
Material | Nitrile |
Powdered | Powder Free |
Finger Thickness (Metric) | 0.13 mm |
Thickness (Metric) | 0.09 mm |
Series | MICROFLEX™ FreeForm™ SE FFS-700 |
FDA 510 (k) | Yes |
Certifications/Compliance (Europe) | EN 374:2003, EN 420:2003 + A1:2009 |
Thickness (English) | 3.5 mil |
Palm Thickness (English) | 3.5 mil |
Latex Free | Yes |
Exterior Finish | Partially Textured |
Certifications/Compliance | ASTM D6319, FDA21 CFR 177-2600 |
AQL | 1.5 |
Palm Thickness (Metric) | 0.09 mm |
Fisherbrand™ Extended Cuff Nitrile Gloves
Choose a glove with exceptional durability and added protection. Fisherbrand™ extended cuff nitrile gloves are extra-thick with textured fingertips for a superior grip and are tested for use with chemotherapy drug exposure.
Disposable | Single-use |
---|---|
Length (Metric) | 300 mm |
Applications | Healthcare,Industrial,Laboratory |
Chemotherapeutic Rated | Yes |
Finger Thickness (English) | 7.5 mil |
Color | Teal |
Length (English) | 11.8 in. |
Quantity | 50 Gloves/Box |
Material | Nitrile |
Powdered | Powder Free |
Sterility | Non-sterile |
Hand | Ambidextrous |
Finger Thickness (Metric) | 0.19 mm |
Thickness (Metric) | Palm: 0.15 mm, Finger: 0.19 mm |
FDA 510 (k) | Yes |
Thickness (English) | Palm: 5.9 mil, Finger: 7.5 mil |
Palm Thickness (English) | 5.9 mil |
Latex Free | Yes |
Exterior Finish | Fingertip Textured |
Certifications/Compliance | ASTM D6319,ASTM D6978,FDA21 CFR 177-2600-US Food Contact Approved,US QSR/FDA 510(k) Medical Examination Grade |
AQL | 1.5 |
Palm Thickness (Metric) | 0.15 mm |
Ansell MICRO-TOUCH™ Nitrafree™ 451 Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Thin, nitrile gloves providing comfortable and reliable protection while supporting breast cancer research in the healthcare setting.
Disposable | Single-use |
---|---|
Length (Metric) | 24 cm |
Applications | Healthcare |
Finger Thickness (English) | 5.5 mil |
Color | Pink |
Length (English) | 9.5 in. |
Material | Nitrile |
Powdered | Powder Free |
Sterility | Non-sterile |
Finger Thickness (Metric) | 0.12 mm |
Thickness (Metric) | 0.08 mm |
Accelerator Free | Yes |
FDA 510 (k) | Yes |
Certifications/Compliance (Europe) | EN 16523-1, EN 374-2 and -4, EN 420:2003 + A1:2009, EN 421:2010, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-1:2016, EN ISO 374-5:2016 |
Product Type | Nitrile Examination Glove |
Thickness (English) | 3.1 mil |
Palm Thickness (English) | 3.1 mil |
Model | MICRO-TOUCH-313016 |
Latex Free | Yes |
Exterior Finish | Fingertip Textured |
Certifications/Compliance | ASTM D6978, ASTM D6319, Category III, CE 0493, FDA21 CFR 177-2600-US Food Contact Approved, ISO 9001, ISO 10993-10:2002, ISO 13485 |
AQL | 1.5 |
Palm Thickness (Metric) | 0.08 mm |
Ansell MICROFLEX™ XCEED™ XC-310 Nitrile Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Ergonomically designed to reduce hand fatigue
Chlorinated | Yes |
---|---|
Length (Metric) | 24 cm |
Chemotherapeutic Rated | Yes |
Finger Thickness (English) | 4.3 mil |
Length (English) | 9.5 in. |
Powdered | Powder Free |
Finger Thickness (Metric) | 0.11 mm |
FDA 510 (k) | Yes |
Thickness (English) | 3 mil |
Palm Thickness (English) | 2.8 mil |
Exterior Finish | Partially Textured |
Certifications/Compliance | ASTM D6319,ASTM D6978,Food Type: Rubber,EN ISO 21420:2020,FDA21 CFR 177-2600-US Food Contact Approved,US QSR/FDA 510(k) Medical Examination Grade |
AQL | 0.65 |
Glove Cuff Style | Beaded |
Tensile Strength | 33 MPa |
Disposable | Single-use |
Applications | Industrial,Healthcare,Laboratory |
Color | Blue |
Material | Nitrile |
Sterility | Non-sterile |
Thickness (Metric) | 0.07 mm |
Accelerator Free | Yes |
Series | MICROFLEX™ XCEED™ XC-310 |
Certifications/Compliance (Europe) | EN 1149-1/2/3 |
Product Type | Nitrile Examination Glove |
Model | XC-310 |
Latex Free | Yes |
Palm Thickness (Metric) | 0.07 mm |
Ansell MICROFLEX™ MidKnight™ Touch 93-732 Nitrile Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Ergonomically designed, thin black nitrile exam gloves provide dexterity along with enhanced comfort fit and feel. Textured fingertips ensure tactility and secure handling of instruments and materials.
Tensile Strength | 18 MPa |
---|---|
Disposable | Single-use |
Length (Metric) | 24.4 cm |
Applications | Healthcare,Industrial,Laboratory |
Chemotherapeutic Rated | No |
Finger Thickness (English) | 3.9 mil |
Color | Black |
Length (English) | 9.6 in. |
Material | Nitrile |
Powdered | Powder Free |
Sterility | Non-sterile |
Finger Thickness (Metric) | 0.10 mm |
Thickness (Metric) | 0.07 mm |
Series | MICROFLEX™ MidKnight™ Touch 93-732 |
FDA 510 (k) | No |
Certifications/Compliance (Europe) | EN 420:2003, A1:2009, EN 421:2010, EN 420:2003, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 16523-1 |
Product Type | Nitrile Examination Glove |
Thickness (English) | 2.8 mil |
Palm Thickness (English) | 2.8 mil |
Latex Free | Yes |
Exterior Finish | Partially Textured |
Certifications/Compliance | EC 1935/2004, Arrêté du 09/04/04, ASTM D6319, EC 2023/2006, FDA21 CFR 177-2600, ISO 13485 |
AQL | 1.5 |
Palm Thickness (Metric) | 0.07 mm |
Fisherbrand™ Powder Free Nitrile Gloves PROMO
Choose a latex-free glove with exceptional durability and puncture resistance. Fisherbrand™ powder-free Nitrile Exam Gloves provide a superior fit, comfort, tactile sensitivity and dexterity.
Disposable | Single-use |
---|---|
Length (Metric) | 24.1 cm |
Finger Thickness (English) | 4.7 mil |
Color | Blue |
Length (English) | 9.5 in. |
Quantity | 100/Box. |
Material | Nitrile |
Powdered | Powder Free |
Finger Thickness (Metric) | 0.12 mm |
Thickness (Metric) | 0.08 mm |
Product Type | Nitrile Examination Glove |
Thickness (English) | 3.5 mil |
Palm Thickness (English) | 3.5 mil |
Latex Free | Yes |
Exterior Finish | Fully Textured |
Certifications/Compliance | ASTM D6319, FDA 21 CFR 177-2600 |
AQL | 1.5 |
Palm Thickness (Metric) | 0.09 mm |
Fisherbrand™ Premium Foam-Insulated Leather Drivers Gloves
Constructed from top grain cowhide leather with foam insulation to keep hands warm during cold applications.
Series | 77-208 |
---|---|
Product Type | Driver Gloves |
Color | Natural |
Style | Gunn |
Material | Top Grain Cowhide Leather |
Grade | Premium |
Liner Material | Foam |
For Use With (Application) | Farming, Construction, Machine Operation, and Refrigeration |
Glove Cuff Style | Open |
Hand | Specific |
Ansell MICROFLEX™ MidKnight™ Black MK-296 Nitrile Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Powder free, nitrile examination gloves with distinct black color hides stains and provides contrast for improved visibility.
Tensile Strength | 27 MPa |
---|---|
Disposable | Single-use |
Length (Metric) | 24.38 cm |
Applications | Healthcare,Laboratory,Industrial |
Chemotherapeutic Rated | No |
Finger Thickness (English) | 5.9 mil |
Color | Black |
Length (English) | 9.6 in. |
Material | Nitrile |
Powdered | Powder Free |
Finger Thickness (Metric) | 0.15 mm |
Thickness (Metric) | 0.12 mm |
Series | MICROFLEX™ MidKnight™ Black MK-296 |
FDA 510 (k) | Yes |
Certifications/Compliance (Europe) | EN 1149-1/2/3, EN 420:2003 + A1:2009 |
Thickness (English) | 4.7 mil |
Palm Thickness (English) | 4.7 mil |
Latex Free | Yes |
Exterior Finish | Fully Textured |
Certifications/Compliance | ASTM D6319, FDA21 CFR 177-2600, ISO 1119 |
AQL | 1.5 |
Palm Thickness (Metric) | 0.12 mm |