Microbiological Specimen Container Guide
Protect Your Specimens
and Your People
Explore specimen collection kits and containers for storing and transporting samples suspected of containing bacteria, viruses, and other microorganisms.
Trusted Industry Leaders
Featured Brands
Fisherbrand
BD
Copan
Puritan
STARplex Scientific
Edge
Need It in a Hurry?
Look for the Fisher Scientific Edge icon to find ready-to-ship products available for standard same-day shipping when ordered by 2 p.m.*
Shop Ready-to-Ship Products Now* Restrictions may apply.
Popular with Customers Like You
Featured Products
Microbiological Specimen Transport and Collection Kits
BD Universal Viral Transport Collection Kits
BD Universal Viral Transport Collection Kits
Thermo Scientific™ MicroTest™ M4RT 3mL w/o Beads NP Swab Kit
Microbiological Specimen Transport and Collection Kits
Thermo Scientific™ MicroTest™ M4RT 3mL w/o Beads NP Swab Kit
Copan Diagnostics UTM™ Bulk Viral Specimen Collection 16 x 100 mm Tube
Microbiological Specimen Transport and Collection Kits
Copan Diagnostics UTM™ Bulk Viral Specimen Collection 16 x 100 mm Tube
Thermo Scientific™ ESwab™
Microbiological Specimen Transport and Collection Kits
Thermo Scientific™ ESwab™
Learn About Key Attributes
Commonly Asked Questions
Specimen Collection Kit
What Is in a
Specimen Collection Kit?
A specimen collection kit contains the elements needed for secure collection, storage, and transport of blood, stool, urine, and other microbiological or biomedical specimens. Depending on the application, these kits should include a swab or applicator sealed in a sterile vial, tube, or pouch. Vials containing transport or other media may also be components of specimen collection kits. Vials should have a screw cap or other secure closure. Because the specimens are subject to degradation, directions for their proper use should be provided and closely followed.
How Do You Use a
Specimen Collection Kit?
Follow the proper procedures for collecting microbiological or biomedical specimens. Begin the process by donning the appropriate masks, gloves, goggles, gowns, and other safety gear to minimize your exposure to infectious diseases and prevent sample contamination.
  1. Open the container
  2. Swab the appropriate area of the patient
  3. Insert the swab into the transport media, breaking the swab at the scored line as needed
  4. Secure the closure
  5. Label the vial with the patient information
  6. Transport the specimen to the lab for processing as soon as possible
Specimen Collection Kit