Autosampler Vial Inserts
Thermo Scientific™ 9mm Autosampler Inserts Available on GSA/VA Contract for Federal Government customers only.
Clear glass or polypropylene inserts for 9mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 160μL to 3.5mL.
Thermo Scientific™ 8mm Autosampler Inserts Available on GSA/VA Contract for Federal Government customers only.
Clear glass or polypropylene inserts for 8mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 125μL to 200μL.
Thermo Scientific™ National Polyspring Vial Inserts Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Polyspring Vial Inserts are available as 150 or 300 μL.
Thermo Scientific™ National Flat-Bottom Vial Inserts Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Flat-Bottom Vial Inserts are bulk-packed for convenience.
Krackeler Scientific Vial Inserts
For microvolume sampling
Thermo Scientific™ SUN-SRi™ StepVial System™ Inserts Available on GSA/VA Contract for Federal Government customers only.
Automatically seat at optimal height and center; no springs or supports needed
Thermo Scientific™ National Scientific Inserts for Target LoVial™ wide-opening vials
With a nominal O.D. of 6mm, these Thermo Scientific™ National Inserts fit Target LoVial Wide-Opening Vials.
Restek™ Glass Vial Inserts
Specially designed for 2.0mL, 11mm crimp-top and 2.0mL, 9mm short-cap, screw-thread vials
Thermo Scientific™ 11mm Autosampler Inserts Available on GSA/VA Contract for Federal Government customers only.
Made of clear glass or polypropylene, these 11mm inserts are available in a variety of configurations with usable volumes from 250μL to 450μL.
Thermo Scientific™ 13mm Autosampler Inserts
Made of clear Type 1 Class A borosilicate glass, these inserts for 13mm HPLC and GC autosampler vialshave usable volumes from 350μL to 850μL.
Thermo Scientific™ National Inserts for Standard-Opening Vials
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
Restek™ Polypropylene Vial Inserts
Specially designed for 2.0mL, 11mm crimp-top and 2.0mL, 9mm short-cap, screw-thread vials.
DWK Life Sciences™ MicroLiter Limited Volume Inserts for 9mm and 11mm Autosampler Vials, Untreated
MicroLiter limited volume inserts are made from Type 1 borosilicate glass or polypropylene and meet strict tolerances. Conical inserts, inserts with plastic springs, and flat bottom inserts are available to assist in consistent sampling of low-volume samples.
DWK Life Sciences™ MicroLiter Limited Volume Inserts for 9mm and 11mm Autosampler Vials, Silanized
MicroLiter limited volume inserts are made from Type 1 borosilicate glass or polypropylene and meet strict tolerances. Conical inserts, inserts with plastic springs, and flat bottom inserts are available to assist in consistent sampling of low-volume samples.
Thermo Scientific™ Chromacol™ GOLD-Grade Inert Vials and Inserts
Ensure sample security and integrity with inert vials that have the highest recovery rates, an ultra-low pH shift, and lowest alkali content.
Thermo Scientific™ National Inserts for 15 × 45mm 4mL Vials
Thermo Scientific™ National Vial Inserts offer a variety of options incuding glass conical, Polyspring and pulled point.
Thermo Scientific™ National Microvolume Inserts for 8 × 40mm Shell Vials
Thermo Scientific™ National Microvolume Inserts are available in pulled point or precision point design to suit various application needs.
Restek™ Glass Vial Inserts for 4.0 mL WISP 48 Screw-Thread Step Vials
Specially designed for 4.0mL, 11mm crimp-top and 4.0mL, 9mm short-cap, screw-thread vials.
  spinner