Dissection Forceps
Fisherbrand™ Hemostatic Forceps Smooth Jaws Pean Str
Fisherbrand™ Pean Hemostatic Forceps are available in an assortment of lengths and jaw types.
Fisherbrand™ Hemostat Rankin-Kelly Forceps Edge
Fisherbrand Hemostat Rankin-Kelly Forceps are primarily used for clamping large blood vessels and manipulating heavy tissue. The multipurpose forceps are popular amongst the fields of healthcare, science and research, education and more! Made from ASTM™ standard (F899-84) surgical stainless steel.
Fisherbrand™ Dissecting Blunt-Pointed Forceps
Fisherbrand Blunt-Pointed Dissection Forceps are constructed from heavy duty stainless-steel, perfect for long lasting use.
Fisherbrand™ Dissecting Cushing Delicate Thumb Forceps
Fisherbrand Dissecting Cushing Delicate Thumb Forceps are made from ASTM™ stainless steel construction. These forceps are light/medium weight and are primarily used to handle delicate tissue.
Thermo Scientific™ Shandon™ Cartilage Curved Thumb Forceps, Curved, Fine Point, standard, 5 in. (12.7cm)
Choose 5″ (12.7cm) cartilage thumb forceps with curved arms and fine points for quality and precision.
Fisherbrand™ Hemostats Kelly Forceps Edge
Fisherbrand Hemostats Kelly Forceps are primarily used to control blood flow in surgical procedures. ASTM™ standard (F899-84) stainless steel.
Fisherbrand™ Dissecting Fine-Pointed Forceps Edge
Fisherbrand Dissecting Fine-Pointed Forceps are used for fine dissection. Fine, serrated tips with guide pins.
Fisherbrand™ Dissecting Tissue Forceps
Fisherbrand Dissecting Dressing Forceps are treated to increase resistance to corrosion. Constructed from ASTM™ standard stainless steel.
Thermo Scientific™ Shandon™ Microdissecting Fine Tips Thumb Forceps, Fine Tips, 5 in. (12.7cm)
Choose 5″ (12.7cm) microdissecting thumb forceps with fine tips for quality and precision.
Thermo Scientific™ Shandon™ Cartilage Straight Thumb Forceps, Straight, Standard, 5 in. (12.7cm)
Choose 5″ (12.7cm) cartilage thumb forceps with straight arms and fine points for quality and precision.
Fisherbrand™ Hemostats Halstead Mosquito Forceps
Fisherbrand Hemostats Halstead Mosquito Forceps are primarily used to control blood flow in surgical procedures. ASTM™ standard (F899-84) stainless steel.
Fisherbrand™ Medium-Pointed Dissecting Forceps
Fisherbrand Medium-Pointed Dissection Forceps are used for grasping tissue and materials during dissection. ASTM™ standard stainless-steel
Fisherbrand™ Dissecting Dressing Forceps Edge
Fisherbrand Dissecting Dressing Forceps are treated to increase resistance to corrosion.
Fisherbrand™ Micro Adson Tissue Forceps 1X2 Teeth German Steel
Micro-Adson tissue forceps are for holding and manipulating delicate tissues.
Fisherbrand™ Dissecting Extra-Fine-Pointed Precision Splinter Forceps
Fisherbrand Dissecting Extra-Fine-Pointed Precision Splinter Forceps are treated for increased corrosion resistance. Made from ASTM™ standard (F899-84) surgical stainless steel.
Thermo Scientific™ Shandon™ Broad Point Dressing Thumb Forceps
Select a fine serrated tip ranging in size from 4.5 to 12 in. (11.4 to 30.5cm) on the Shandon™ Broad Point Dressing Thumb Forceps.
Fisherbrand™ Rochester-Pean Serrated Forceps
Fisherbrand Rochester-Pean Serrated Forceps are primarily designed for hemostasis by clamping of larger vessels. The multipurpose forceps are popular amongst the fields of healthcare, science and research, education and more! Made from ASTM™ standard (F899-84) surgical stainless steel.
Thermo Scientific™ Shandon™ Dura Strip™ Forceps, Serrated, heavy-duty, standard, 8.5 in. (21.6cm)
Choose 8.5″ (21.6cm) heavy-duty, serrated Dura Strip Forceps for quality and precision
Fisherbrand™ Curved, Serrated or Smooth Tip Microforceps
Fisherbrand Curved, Serrated or Smooth Tip Microforceps are designed for high precision work. Made from ASTM™ standard (F899-84) stainless steel.
Fisherbrand™ Premium Tissue Forceps 1X2 Teeth 5 in. German Steel
Tissue forceps are specially designed to grasp tissue with minimal damage to the organic material.
Fisherbrand™ Filter Forceps PTFE Coated
Easy, effortless handling of membranes. PTFE coated to prevent sticking and corrosion.
Fisherbrand™ Hemostatic Pean Forceps Serrated
Fisherbrand™ Pean Hemostatic Forceps are available in an assortment of lengths and jaw types.
Fisherbrand™ Premium Adson Dressing Forceps 4.75 in. German Steel
Premium adson dressing forceps are for holding and manipulating delicate tissues.
Graham-Field™ Kelly Hemostatic Forceps
For medical and surgical procedures
Thermo Scientific™ Kuehne Coverglass Thumb Forceps, 4 in. (10.2cm)
Choose 4″ (10.2cm) Kuehne Coverglass Thumb Forceps, featuring angled tips for quality and precision.
Thermo Scientific™ Shandon™ Microdissecting Extra Fine Tip Thumb Forceps, Extra Fine Tips, curved, 4.5 in. (11.4cm)
Choose 4.5″ (11.4cm) microdissecting extra fine tip thumb forceps with extra fine tips and curved arms for quality and precision.
Fisherbrand™ Micro Jewelers Forceps PTFE-Coated
Fisherbrand™ Micro-Jewelers Dissecting Forceps are stainless steel with superfine tips coated with PTFE for non-sticking applications
Fisherbrand™ Self-Closing Dissecting Forceps
Blunt-pointed tips of the Fisherbrand™ Self-Closing Forceps are closed in the resting state. Useful for clamping applications. Multipurpose. Made from ASTM™ standard (F899-84) surgical stainless steel.
  spinner