Dye Protein Labeling Reagents
BD 7-AAD Staining Solution
Convenient, ready-to-use nucleic acid dye that can be used in place of propidium iodide (PI)
GE Healthcare Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester) Available on GSA/VA Contract for Federal Government customers only.
Use for highly accurate labeling of amine groups. GE Healthcare Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester) contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.
Invitrogen™ eBioscience™ 7-AAD Viability Staining Solution Non-distribution product as customer accommodation.
Ready-to-use solution for the exclusion of nonviable cells in flow cytometric analysis. Can be used in combination with PE- and FITC-conjugated antibodies in 2-color analysis. The advantage of 7-AAD over PI is that there is minimal spectral overlap between these emissions.
Molecular Probes™ Biotin Azide (PEG4 carboxamide-6-Azidohexanyl Biotin) Non-distribution product as customer accommodation.
biotin azide (PEG4 carboxamide-6-azidohexanyl biotin)
Molecular Probes™ Biotin-XX, SE (6-((6-((Biotinoyl)Amino)Hexanoyl)Amino)Hexanoic Acid, Succinimidyl Ester) Non-distribution product as customer accommodation.
Biotin-XX, SE (6-((6-((Biotinoyl)Amino)Hexanoyl)Amino)Hexanoic Acid, Succinimidyl Ester)
Molecular Probes™ Biotin Alkyne (PEG4 carboxamide-Propargyl Biotin) Non-distribution product as customer accommodation.
biotin alkyne (PEG4 carboxamide-propargyl biotin)
GE Healthcare Amersham™ CyDye™ Antibody Labeling Kits Available on GSA/VA Contract for Federal Government customers only.
Convenient way to tag antibodies with CyDye fluorescent dyes
GE Healthcare Amersham™ CyDye™ Reactive Dye 5-Packs Available on GSA/VA Contract for Federal Government customers only.
Use for the most accurate labeling. GE Healthcare Amersham™ CyDye™ Reactive Dye 5-Packs contain derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups.
Novus Biologicals™ InstantBlue Protein Stain
InstantBlue is a ready to use Coomassie protein stain for polyacrylamide gels.
Molecular Probes™ 4-Acetamido-4'-Maleimidylstilbene-2,2'-Disulfonic Acid, Disodium Salt Non-distribution product as customer accommodation.
4-acetamido-4'-maleimidylstilbene-2,2'-disulfonic acid, disodium salt
Invitrogen™ eBioscience™ Propidium Iodide Staining Solution Non-distribution product as customer accommodation.
Ready-to-use solution for the exclusion of nonviable cells in flow cytometric analysis. Propidium Iodide binds to double-stranded DNA by intercalating between basepairs, but is excluded from cells with intact plasma membranes.
Abnova™ Fluorescent Dye 488-I Phalloidin
Fluorescence Dye 488-I with Phalloidin conjugate
GE Healthcare Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester) Available on GSA/VA Contract for Federal Government customers only.
Suitable for general labeling of amine residues
GE Healthcare Cy3 NHS Ester Mono-Reactive CyDye Available on GSA/VA Contract for Federal Government customers only.
Use for highly accurate labeling of amine groups. GE Healthcare Cy3 NHS Ester Mono-Reactive CyDye contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.
Abnova™ Fluorescent Dye 647-I Phalloidin
Fluorescence Dye 647-I with Phalloidin conjugate
GE Healthcare Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide) Available on GSA/VA Contract for Federal Government customers only.
For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides
Molecular Probes™ DTNB; Ellman's Reagent, 5,5'-Dithiobis-(2-Nitrobenzoic Acid) Non-distribution product as customer accommodation.
DTNB; Ellman's Reagent, 5,5'-Dithiobis-(2-Nitrobenzoic Acid)
GE Healthcare Cy5.5 NHS Ester Mono-Reactive CyDye Available on GSA/VA Contract for Federal Government customers only.
Use for highly accurate labeling of amine groups. GE Healthcare Cy5.5 NHS Ester Mono-Reactive CyDye contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.
Abnova™ Fluorescent Dye 405-I Phalloidin
Fluorescence Dye 405-I with Phalloidin conjugate
Molecular Probes™ QSY™ 21 Carboxylic Acid, Succinimidyl Ester Non-distribution product as customer accommodation.
QSY™ 21 carboxylic acid, succinimidyl ester
GE Healthcare Amersham™ QuickStain Protein Labeling Kit
Amersham QuickStain enables protein detection directly after SDS-PAGE or transfer. Supports Western blot normalization for quantitative Western blotting.
Molecular Probes™ QSY™ 9 Carboxylic Acid, Succinimidyl Ester Non-distribution product as customer accommodation.
QSY™ 9 carboxylic acid, succinimidyl ester
Molecular Probes™ QSY™ 7 Carboxylic Acid, Succinimidyl Ester Non-distribution product as customer accommodation.
QSY™ 7 carboxylic acid, succinimidyl ester
GE Healthcare DeStreak Rehydration Solution Available on GSA/VA Contract for Federal Government customers only.
Highly suitable for both analytical and preparative gels. DeStreak Rehydration Solution improves reproducibility and quality of 2-D gels by preventing streaking.
Abnova™ Fluorescent Dye 488-I Streptavidin
Fluorescence Dye 488-I with Streptavidin conjugate
Abnova™ Fluorescent Dye 594-I Phalloidin
Fluorescence Dye 594-I with Phalloidin conjugate
Molecular Probes™ Azido, PEO (4 propionic Acid, Succinimidyl Ester (3-(Azidotetra(Ethyleneoxy))Propionic Acid, Succinimidyl Ester)) Non-distribution product as customer accommodation.
Azido, PEO (4 propionic Acid, Succinimidyl Ester (3-(Azidotetra(Ethyleneoxy))Propionic Acid, Succinimidyl Ester))
Molecular Probes™ Biotin-XX, SSE (6-((6-((Biotinoyl)Amino)Hexanoyl)amino)Hexanoic Acid, Sulfosuccinimidyl Ester, Sodium Salt) Non-distribution product as customer accommodation.
Biotin-XX, SSE (6-((6-((Biotinoyl)Amino)Hexanoyl)amino)Hexanoic Acid, Sulfosuccinimidyl Ester, Sodium Salt)
Novus Biologicals™ JC-1 mitochondrial membrane potential dependent fluorescent dye
Mitochondrial membrane potential dependent fluorescent dye.
  spinner