Kuderna Danish Devices
DWK Life Sciences Kimble™ Kontes™ 10mL SLOW-DRY™ Concentrator Tube for Evaporative Concentrators
The SLOW-DRY™ concentrator tube reduces the risk of concentrating a sample to dryness.
DWK Life Sciences Kimble™ Kontes™ 10mL Concentrator Tube with Hooks for Evaporative Concentrators Available on GSA/VA Contract for Federal Government customers only.
This concentrator tube, for use with Kuderna-Danish and other concentrators, may be detached from the complete assembly and stoppered for analysis or storage.
PYREX™ Kuderna-Danish Replacement Flasks Available on GSA/VA Contract for Federal Government customers only.
Includes pair of 1.25 in. stainless-steel springs and two sizes of Keck clamps
PYREX™ Replacement Tube for Kuderna-Danish Apparatus Available on GSA/VA Contract for Federal Government customers only.
Replacement tube for use with Kuderna-Danish apparatus
PYREX™ Kuderna-Danish Concentrator Apparatus With Hooks
Complete 1L PYREX Kuderna-Danish concentrator apparatus
DWK Life Sciences Wheaton™ Evaporative Flask Available on GSA/VA Contract for Federal Government customers only.
For concentration of trace amounts of analyte in organic solvent
DWK Life Sciences Kimble™ Kontes™ Kuderna-Danish Apparatus with a Graduated Concentrator Tube
The Kuderna-Danish apparatus was developed for the concentration of trace amounts of sample dissolved in organic solvents. The column is designed to speed evaporation with reduced hold-up. It is very useful in sample preparation before analysis with solvents such as petroleum ether or hexane.
DWK Life Sciences Wheaton™ Three-ball Snyder Column Available on GSA/VA Contract for Federal Government customers only.
For concentration of trace amounts of analyte in organic solvent
DWK Life Sciences Kimble™ Kontes™ 10mL Tapered Concentrator Tube for Evaporative Concentrators Available on GSA/VA Contract for Federal Government customers only.
This tapered concentrator tube, for use with Kuderna-Danish and other concentrators, may be detached from the complete assembly and stoppered for analysis or storage.
DWK Life Sciences Wheaton™ Kuderna-Danish Evaporative Concentrator With Clear-Seal™ Joints Available on GSA/VA Contract for Federal Government customers only.
For concentration of trace amounts of analyte in organic solvent
DWK Life Sciences Kimble™ Kontes™ 25mL Concentrator Tube for Evaporative Concentrators
This concentrator tube, for use with Kuderna-Danish and other concentrators, may be detached from the complete assembly and stoppered for analysis or storage.
  spinner