Filtered Search Results
Agilent BioTek Synergy LX Multi-Mode Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek Synergy LX multi-mode reader automates many common microplate assays. NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately). For Research Use Only. Not for use in diagnostic procedures.
Width (English) | 15 in. |
---|---|
Shaking | Linear, Orbital, Double Orbital |
Absorbance Range | 200 to 900 nm |
Light Source | Xenon Flash |
For Use With (Application) | Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability |
Width (Metric) | 38.1 cm |
USB Connections | 2 |
Detector Type | Photodiode,PMT |
Reading Options | End Point,Kinetic,Area Scanning,Absorbance Spectral Scanning |
Filters | Monochromator Bandpass |
Accuracy | Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD |
Voltage | 100/240 V |
Bandwidth | ≤5 nm |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Weight (Metric) | 12.3 kg |
Warranty | 1 year |
Depth (English) | 15 in. |
Software Language Versions | English |
Sensitivity | Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP |
Product Type | Multi-mode Microplate Reader |
Linearity | <1% from 0 to 2.5 OD |
Depth (Metric) | 38.1 cm |
Measurement Speed | 24 sec. (96 wells), 76 sec. (384 wells) |
Frequency | 50/60 Hz |
Weight (English) | 27 lb. |
Power Consumption | 24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.) |
Resolution | 0.001 OD |
Agilent BioTek 800 TS Absorbance Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For Research Use Only. Not for use in diagnostic procedures.
Detector Type | Photodiode |
---|---|
Reading Options | Endpoint, Kinetic, Well-area Scanning |
Shaking | Linear (except NB configurations) |
Product Type | Microplate Reader |
Voltage | 100/240 V |
Bandwidth | 10 nm |
Software Type | Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) |
Weight (English) | 18.5 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
For Use With (Application) | Measuring absorbance in microplate-based assays |
Light Source | Tungsten Halogen Lamp |
Interface | USB Type B, USB Type A |
Weight (Metric) | 8 kg |
Agilent BioTek Cytation 5 Cell Imaging Multi-Mode Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. For Research Use Only. Not for use in diagnostic procedures.
Height (Metric) | 44.5 cm |
---|---|
Depth (English) | 20.2 in. |
Width (English) | 16.4 in. |
Contrast Methods | Fluorescence, phase contrast, brightfield, high contrast brightfield, color brightfield and phase contrast |
Shaking | Linear, Orbital, Double Orbital |
Wattage | 250 W max. |
Objectives | 1.25x, 2.5x (2.25x eff), 2.5x (2.75x eff), 4x, 10x, 20x, 40x, 60x |
Absorbance Range | 230 to 999 nm, 1 nm increments |
Light Source | Xenon Flash; 100 mW 680 nm laser (Alpha Detection) |
Width (Metric) | 41.6 cm |
Dynamic Range | Absorbance: 0 to 4 OD; Fluorescence Intensity: 7 decades; Luminesence: >6 decades |
Reading Options | End point, kinetic, spectral scanning and well area scanning |
Accuracy | Monochromator: ±2 nm; Dispenser: ±1 μL or 2% |
Type | Cell Imaging Multi-Mode Microplate Reader |
No. of Dispensers | 2 Syringe Pumps |
Dispense Volume | 5 to 1000 μL, 1 μL increments |
For Use With (Equipment) | BioStack and Third Party Automation Compatible; BioSpa 8 Automated Incubator Compatible |
Depth (Metric) | 51.4 cm |
Weight (English) | 80 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Camera | Sony CMOS, 16-bit Gray Scale, Standard or WFOV |
Temperature Range (Metric) | 4-Zone incubation to 65°C with Condensation Control (±0.2°C at 37°C) |
CO2 Concentration Range | 0 to 20% |
Weight (Metric) | 36.3 kg |
Height (English) | 17.5 in. |
Agilent BioTek MultiFlo FX Multi-Mode Dispenser, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates. For In Vitro Diagnostic Use.
Height (Metric) | 20.3 cm |
---|---|
Speed | 96 wells, 5 μL cass, 10 μL/ well: 3 s, 96 wells, 5 μL cass, 20 μL/ well: 3.5 s, 384 wells, 5 μL cass, 5 μL/ well: 6.5 s, 384 wells, 10 μL cass, 10 μL/ well: 8 s, 384 wells, 1 μL cass, 1 μL/ well: 6 s, 1536 wells, 1 μL cass, 1 μL/ well: 21 s |
Electrical Requirements | 90 W max. |
Depth (English) | 11.8 in. |
Width (English) | 17.2 in. |
For Use With (Application) | Primary/secondary screening assays, compound storage, genomics and proteomics research, ELISA, cell based washing, fixing, staining, volume or concentration normalization |
Width (Metric) | 43.5 cm |
Accuracy | ±5% at 1 μL, ±2.0% at 5 μL, ±2.0% at 10 μL |
Precision | <5% CV at 1 μL, <2.5% CV at 5 μL, <2.0% CV at 10 μL |
For Use With (Equipment) | BioStack and third party automation compatible; BioSpa 8 Automated Incubator compatible |
Product Type | Microplate Dispenser |
Depth (Metric) | 29.2 cm |
Voltage | 100/240 V |
Weight (English) | 14.3 lb. |
Software | LHC2 Software; LHC2 Secure for 21 CFR Part 11 compliance (option); SiLA Compliant driver (option) |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Interface | Color touch screen; Create, edit or run multiple protocols onboard |
Weight (Metric) | 6.5 kg |
Height (English) | 8 in. |
Plate Type | Dispensing (peri pump and syringe): 96-, 384- and 1536-well standard, deep and PCR plates; 6-. 12-, 24- and 48-well plates (dispense tip configurable); Washing: 96-. 384-well standard plates; 6-, 12-, 24-,and 48-well plates (with compatible manifold configuration) |
Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For In Vitro Diagnostic Use.
Height (Metric) | 41 cm |
---|---|
Depth (English) | 20.7 in. |
Width (English) | 15.4 in. |
Shaking | Linear, Orbital, Double Orbital |
Software Type | Gen5 Data Analysis Software,Gen5 Secure for 21 CFR Part 11 compliance (option) |
Absorbance Range | 230 to 999 nm, 1 nm increments |
Incubator Temperature | ±0.2°C at 37°C |
Light Source | Xenon Flash |
For Use With (Application) | HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP |
Width (Metric) | 39 cm |
Dynamic Range | 0 to 4 OD |
Detector Type | Photodiode,Fluorescence/Alpha: Dual PMT or Single PMT (option) |
Accuracy | Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD |
Dispense Volume | 5 to 1000 μL, 1 μL increments |
Product Type | Multi-Mode Microplate Reader |
Linearity | ≤1% from 0 to 3 OD |
Depth (Metric) | 52.5 cm |
Weight (English) | 78 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Power Consumption | 250 W max. |
CO2 Concentration Range | 0 to 20% |
Weight (Metric) | 35 kg |
Resolution | 0.0001 OD |
Height (English) | 16.1 in. |
Agilent BioTek BioSpa 8 Automated Incubator
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. For Research Use Only. Not for use in diagnostic procedures.
Height (Metric) | 48 cm |
---|---|
Depth (English) | 20.1 in. |
Width (English) | 27.2 in. |
Product Type | Automated Microplate Incubator |
Depth (Metric) | 51 cm |
Humidity | 80 to 95% |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Width (Metric) | 69 cm |
Operating Temperature Range (Metric) | Ambient to 45°C |
Height (English) | 18.9 in. |
Agilent BioTek 406 FX Washer Dispenser
The 406 FX is a compact instrument offering fast, full plate washing along with up to six reagent dispensers. For Research Use Only.
Agilent BioTek 405 TS Touch Microplate Washer, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. For In Vitro Diagnostic Use.
Connections | USB (2) |
---|---|
Width (English) | 14 in. |
For Use With (Application) | ELISA, MSD assays, HCS immune cytochemistry, FLIPR Ca2+ flux, Cell-based assays, Magnetic and polystyrene bead processing (gene expression assays, cytokine assays), ELISPOT assays |
Width (Metric) | 35.6 cm |
Assays | ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes |
Flow Rate | High to Low Flow; Optimized rates for cell assays |
Bottles | Optional (4 or 10 L) |
Wash Cycles | 1 to 250 |
Voltage | 100/240 V |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Height (English) | 10 in. |
Weight (Metric) | With internal buffer switching - 16.5 kg; Without - 13.5 kg |
Wash Volume | 25 to 3000 μL/well |
Height (Metric) | 25.4 cm |
Depth (English) | 17 in. |
Fluid Delivery Type | Internal positive displacement pump |
Automation Interface | High Resolution LED Backlit Touch Screen |
Type | Microplate Washer |
Washing Speed | 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s |
Dispense Volume | 25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold) |
Precision | <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) |
Depth (Metric) | 43.2 cm |
Soak Time | Programmable in minutes and seconds, up to 60 min. |
Frequency | 50/60 Hz |
Residual Volume | ≤2 μL/well |
Weight (English) | With internal buffer switching - 36 lb.; Without - 30 lb. |
Display | LED |
Plate Type | 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates |
Agilent BioTek ELx405 Select Deep Well Microplate Washer
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates.
Height (Metric) | 25.4 cm |
---|---|
Depth (English) | 17 in. |
Fluid Delivery Type | One positive displacement pump |
Width (English) | 14 in. |
Temperature (English) Operating | 59°F to 86°F |
Width (Metric) | 35.6 cm |
Type | Microplate Washer |
Precision | ≤3% CV |
Depth (Metric) | 43.2 cm |
Wash Cycles | 1 to 10 |
Voltage | 100/240 V |
Humidity | 10 to 80%, noncondensing |
Soak Time | Programmable in minutes and seconds, up to 60 min. |
Amperage | <8 A |
Frequency | 50/60 Hz |
Residual Volume | ≤2 μL/well |
Weight (English) | 30 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Temperature (Metric) Operating | 15°C to 30°C |
Height (English) | 10 in. |
Display | Backlit LCD |
Weight (Metric) | 13.5 kg |
Plate Type | 96-well Plate, 384-well Plate |
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For In Vitro Diagnostic Use.
Detector Type | Photodiode |
---|---|
Accuracy | ±2 nm |
Measurement Speed | 96 wells: 8 sec., 384 wells: 14 sec. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
For Use With (Application) | Nucleic acid and protein quantification to ELISA and microbial growth assays |
Light Source | Xenon Flash Lamp |
Hertz | 50/60 Hz |
Wavelength Range | 200 to 999 nm, 1 nm increments |
Height (English) | 22 in. |
Agilent BioTek Lionheart LX Automated Microscope
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Lionheart FX automated microscope is a compact system for a broad range of imaging workflows. For Research Use Only. Not for use in diagnostic procedures.
Agilent BioTek Cytation 5 Multi-Mode Only Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. For Research Use Only. Not for use in diagnostic procedures.
Height (Metric) | 44.5 cm |
---|---|
Depth (English) | 20.2 in. |
Width (English) | 16.4 in. |
Shaking | Linear, Orbital, Double Orbital |
Wattage | 250 W max. |
Absorbance Range | 230 to 999 nm, 1 nm increments |
Light Source | Xenon Flash; 100 mW 680 nm laser (Alpha Detection) |
Width (Metric) | 41.6 cm |
Dynamic Range | Absorbance: 0 to 4 OD; Fluorescence Intensity: 7 decades; Luminesence: >6 decades |
Reading Options | End point, kinetic, spectral scanning and well area scanning |
Accuracy | Monochromator: ±2 nm; Dispenser: ±1 μL or 2% |
Type | Multi-Mode Only Microplate Reader |
No. of Dispensers | 2 Syringe Pumps |
Dispense Volume | 5 to 1000 μL, 1 μL increments |
For Use With (Equipment) | BioStack and Third Party Automation Compatible; BioSpa 8 Automated Incubator Compatible |
Depth (Metric) | 51.4 cm |
Weight (English) | 80 lb. |
Software | Gen5 Microplate Reader and Imager Software included; Gen5 Image+ and Image Prime software available for full image analysis; Gen5 Secure for 21 CFR Part 11 compliance (option) |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Temperature Range (Metric) | 4-Zone incubation to 65°C with Condensation Control (±0.2°C at 37°C) |
CO2 Concentration Range | 0 to 20% |
Weight (Metric) | 36.3 kg |
Height (English) | 17.5 in. |
Agilent BioTek Epoch 2 Microplate Spectrophotometer
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For Research Use Only. Not for use in diagnostic procedures.
Detector Type | Photodiode |
---|---|
Accuracy | ±2 nm |
Measurement Speed | 96 wells: 8 sec., 384 wells: 14 sec. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
For Use With (Application) | Nucleic acid and protein quantification to ELISA and microbial growth assays |
Light Source | Xenon Flash Lamp |
Hertz | 50/60 Hz |
Wavelength Range | 200 to 999 nm, 1 nm increments |
Height (English) | 22 in. |
Agilent BioTek Epoch Microplate Spectrophotometer, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Epoch microplate spectrophotometer has monochromator-based optics, for a filter-free, wide wavelength range for UV-Vis absorbance measurements. For In Vitro Diagnostic Use.
Height (Metric) | 19.6 cm |
---|---|
Cuvette Size | 0.5 mm |
Width (English) | 12 in. |
Software Type | Gen5 Data Analysis Software |
Absorbance Range | 200 to 999 nm, 1 nm increments |
Light Source | Xenon Flash Lamp |
Width (Metric) | 30.5 cm |
Accuracy | ±2 nm |
Precision | ±0.2 nm |
Product Type | Microplate Reader |
Voltage | 100/240 V |
Bandwidth | 5 nm |
Weight (English) | <15 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Hertz | 50/60 Hz |
Wavelength Range | 200 to 999 nm |
Weight (Metric) | 6.8 kg |
Resolution | 0.0001 OD |
Height (English) | 7.7 in. |
Plate Type | 6 to 384-well Plates |
Agilent BioTek ELx405 Select Microplate Washer, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. For In Vitro Diagnostic Use.
Height (Metric) | 25.4 cm |
---|---|
Depth (English) | 17 in. |
Fluid Delivery Type | One positive displacement pump |
Width (English) | 14 in. |
Temperature (English) Operating | 59°F to 86°F |
Width (Metric) | 35.6 cm |
Type | Microplate Washer |
Precision | ≤3% CV |
Depth (Metric) | 43.2 cm |
Wash Cycles | 1 to 10 |
Voltage | 100/240 V |
Humidity | 10 to 80%, noncondensing |
Soak Time | Programmable in minutes and seconds, up to 60 min. |
Amperage | <8 A |
Frequency | 50/60 Hz |
Residual Volume | ≤2 μL/well |
Weight (English) | 30 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Temperature (Metric) Operating | 15°C to 30°C |
Height (English) | 10 in. |
Display | Backlit LCD |
Weight (Metric) | 13.5 kg |