UserName
Viewing profile as user:
Chemical Stockroom
Chemical Stockroom
Welcome, Chemical Stockroom Managers!
As the stockroom manager, you are charged with the safe handling of hazardous chemicals every single day. Your responsibilities are far-reaching, extending beyond the storeroom walls to lab personnel, building staff, campus residents and the environment. That's why the Fisher Scientific channel is pleased to offer these tools just for you. Here, you'll find storage recommendations, chemical compatibility charts and a look at our most popular product essentials to accommodate various research applications undertaken by your institution.
We Have What You Need to Stock Up
Fisher Chemical Stockroom Essentials for Research and Productivity
Chemicals should react in the lab, not in the stockroom. The inadvertent mixing of inventory can produce toxic vapor/gas, fire or explosion. Stay safe in the storeroom; adhere to the following prescribed precautions and consult the chemical compatibility tables (below) for caustic combinations. For product-specific information, refer to the Safety Data Sheet (SDS) search tool.
To stay competitive in scientific research, pharmaceuticals and other chemical production fields, you need the best reagents. The Fisher Chemical™ brand has you covered.
The link below will take you to solvents in six application-specific grades, bioreagents for all phases of biochemistry, and fine chemicals packaged for protection from air and moisture.
For reference, this information can be found on page 11 of the Chemical Stockroom Handbook.
Chemical Stockroom General Guidelines
General Guidelines
  • Protect eyes and skin: lab safety glasses with side shields, lab coats and closed-toe shoes must be worn for basic personal protection
  • Safely space shelves and racks to accommodate the upright removal of the largest chemical container; prevent tipping and dripping with adequate clearance
  • Identify and substitute safer chemical alternatives
  • Keep hazardous materials away from heat and direct sunlight to prevent the degradation of chemicals and deterioration of storage containers and labels
  • Do not store hazardous materials (except cleaners) under sinks
  • Avoid chemical stockpiling; procure hazardous materials as needed
  • Limit fume hood storage of hazardous materials
  • Conduct periodic cleanouts to minimize accumulation of chemicals
  • Keep all food (including gum), beverages, tobacco and open cosmetics outside the work area
Other Resources
Fisherbrand™ Products
More than 10,000 budget-friendly products built on the guiding principle to provide quality, reliability and value.
Browse Fisherbrand Lab Essentials
Globally Harmonized System
When it comes to chemical hazards, are you protecting what matters most in your facility? The Fisher Scientific channel can help.
Learn More
Acids and Bases
Isolate acids:
  • From reactive metals, including sodium, potassium and magnesium
  • From sodium cyanide, iron sulfide, calcium carbide and other compounds that can react to produce toxic fumes/gases
  • Place combustible organic carboxylic acids (i.e., acetic acid) in a flammable storage locker; store inorganic acids in acid storage cabinets
  • Store acids and bases in air-tight containers with snug-fitting caps; avoid loose lids or glass stoppers; use vented caps when necessary to prevent over-pressurization
  • Keep piranha etch and aqua regia in a fume hood at all times
  • Use non-aluminum drip trays for aqueous sodium and potassium hydroxide solutions; isolate nitric acid when utilizing secondary containment
  • Safely transfer containers of acid and base solutions using bottle carriers
  • Never pour water into acid; slowly add the acid to the water and stir
Flammable and Combustible Liquids
  • Store flammable and combustible liquids away from oxidizers and heat producers
  • House flammable and combustible liquids in excess of 10 gallons (per room) in approved flammable storage cabinets (under the hood or stand-alone); limit liquids in secondary containers (i.e., squeeze bottles) to 10 gallons or less
    • Adhere to OSHA regulations for safe storage: 60 gallons of Class I and/or Class II liquids or 120 gallons of Class III liquids per cabinet; Class I liquids cannot be stored in a basement or pit without an approved ventilation system
    • Use only approved and well-labeled refrigerators and freezers for storing flammable liquids; never store lunch with science
    Hazard Information
    GHS = Globally Harmonized System of Classification and Labeling
    Created by the UN, the GHS was designed to create a universal standard that would replace the multiple and diverse chemical classifications systems used across the globe. OSHA has revised its Hazard Communication Standard (HCS) to align with the GHS, which uses pictogram symbols to represent the different types of hazards.
    Learn More About GHS Hazard Symbols
    NFPA Hazard Code Ratings
    The National Fire Protection Association (NFPA) has developed a numerical rating system that reflects the health, flammability, self-reactivity and other hazards of materials, including reaction with water, as specified in NFPA 704, Standard System for the Identification of the Hazards of Materials for Emergency Response. Potential hazards are evaluated based on the degree of hazard, and the numerical rating is placed inside the universal NFPA symbol.
    Learn More About Hazard Codes
    ChemAlert Guide
    The ChemAlert Storage Guide is a color-coded system that provides information about storage requirements based on various safety factors. The recommended ChemAlert storage color (category) is shown on the product label and matches the band at the bottom of the FisherLOCK™ cap. Combine the ChemAlert guide, NFPA and GHS indicators, and the product SDS (scanned from the QR code) to create a complete profile of critical safety information for chemical handling and storage.
    The storage code is also spelled out for clarification:
    Red (R): Flammable. Store in area segregated for flammable reagents.
    Red (R): Flammable. Store in area segregated for flammable reagents.
    White (W): Corrosive. May harm skin, eyes, mucous membrane. Store away from red-, yellow- and blue-coded reagents.
    Red (R): Flammable. Store in area segregated for flammable reagents.
    Blue (B): Health hazard. Toxic if inhaled, ingested or absorbed through skin. Store in secure area.
    Red (R): Flammable. Store in area segregated for flammable reagents.
    Gray (G): Presents no more than moderate hazard in any of the categories above. For general chemical storage.
    Red (R): Flammable. Store in area segregated for flammable reagents.
    Yellow (Y): Reactive and oxidizing reagents. May react violently with air, water or other substances. Store away from flammable and combustible materials.
    Yellow (Y): Reactive and oxidizing reagents. May react violently with air, water or other substances. Store away from flammable and combustible materials.
    Exception: Reagent incompatible with other reagents of the same color bar. Store separately.
    NOTE: ChemAlert is a guide only. It should be supplemented by reading the rest of the label (which provides detailed instructions in the event of accidental exposure, spill or fire, and applicable GHS, OSHA, DOT and ANSI* data), the appropriate Safety Data Sheet (SDS) and standard references.
    Packaging
    Compliant with all applicable government regulations, our packaging solutions are designed to keep you safe, preserve the integrity of your chemicals, protect the environment, and provide convenient handling and storage.
    Designed for Your Safety
    Safety Info via QR Codes
    React quickly to safety issues with Fisher Chemical QR codes. Scan the QR code on the label with a smartphone or tablet for instant access to product SDS and CofA.
    Safety Info via QR Codes
    Safe-Cote™ Protects You and Your Chemicals
    Used for certain solvents and acids, these plastic-coated glass bottles can temporarily contain residual chemical and broken glass if the bottle breaks accidentally, protecting employees and the laboratory environment.
    Safe-Cote™ Protects You and Your Chemicals
    Resources
    Within the handbook, you’ll find storage recommendations, chemical compatibility charts and recommended essential products to stock to accommodate various research applications.
    Filename
    fisher-chemical-chemical-stockroom-handbook-guide.pdf
    Size
    5 MB
    Format
    application/pdf
    Review the numerical rating system that reflects the health, flammability, self-reactivity and other hazards of materials.
    Filename
    fisher-chemcial-nfpa-hazard-code-flyer.pdf
    Size
    945 KB
    Format
    application/pdf
    Need More Resources?
    Explore our Chemical Resource Center for videos and more information on our overall portfolio.
    Our Brands