Get a Microvolume Plate with Agilent BioTek Epoch 2 Purchase
When you purchase a qualifying Agilent BioTek Epoch 2 Microplate Spectrophotometer, you’ll get a Take3 Microvolume Plate at no additional cost. To take advantage of this offer, order using the catalog numbers below, log in to the Redemption Center, and complete the redemption form to receive your microvolume plate.
NOTE: Promotion may be used for up to two qualifying products purchased on the same order.
Features and Benefits
The Agilent BioTek Epoch 2 Microplate Spectrophotometer performs UV-Vis absorbance measurements in 6- to 384-well microplates, cuvettes, and in microvolume samples. The Take3 Microvolume Plate lets you decrease sample size to a 2 μL volume. Each Take3 plate includes spaces for 16 x 2 μL microspots, one standard 1 cm cuvette, and two BioCells.
Catalog Number | Description | ||
---|---|---|---|
Agilent BioTek Take3 Microvolume Plate | Learn More |
Restrictions
Offer valid from July 1, 2023 to June 30, 2024. Redemptions must be received by June 30, 2025.
Redeemed product value is based on Fisher Scientific list price. The total value of redemption items cannot exceed the maximum redeemable threshold of the promotion. Offer can be redeemed one time per new laboratory. Offer cannot be combined with any other offers. Orders cannot be combined to qualify unless otherwise stated. Each qualifying item can only count toward one promotion redemption.
Promotion may be used for up to two qualifying products purchased on the same order.
This promotion is not available to our current or potential healthcare customers. Healthcare customers include any healthcare practitioner, other healthcare provider or any individual or organization authorized to prescribe, dispense, purchase or influence the acquisition or use of medical devices or supplies for clinical use. Healthcare customers include, without limitation, any “Health Care Practitioner” under the Massachusetts Marketing Code of Conduct Law (defined at 105 C.M.R §907.004) or a “Health Care Provider” under the Vermont Prescribed Products Gift Ban and Disclosure Law (defined at Vt. Stat. Ann. tit. 18, §4631a(8)). By participating in this promotion, you warrant that you are not a current or potential healthcare customer. Thermo Fisher Scientific reserves the right to determine your status in the event of a dispute.