Filtered Search Results
Kimtech™ Pure G3 Sterile Latex Gloves
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Sterile, hand-specific latex gloves are suitable for double-donning and provide sensitive protection. Kimtech™ Pure G3 Sterile Latex Gloves recommended for Class 10/ISO 4 or higher cleanrooms.
| Tensile Strength | 24 MPa |
|---|---|
| Laboratory/Cleanroom Use | Controlled Environments |
| Disposable | Single-use |
| Length (Metric) | 30.5 cm |
| Finger Thickness (English) | 8.7 mil |
| Color | Natural |
| Length (English) | 12 in. |
| Material | Latex |
| Powdered | Powder Free |
| Sterility | Sterile |
| Hand | Specific |
| Finger Thickness (Metric) | 0.22 mm |
| Thickness (Metric) | 0.20 mm |
| Accelerator Free | Yes |
| Series | Kimtech™ Pure G3 |
| Certifications/Compliance (Europe) | EN ISO 374-1:2016, EN 374-4:2003, EN ISO 374-5:2016 |
| Product Type | Cleanroom Gloves |
| Thickness (English) | 8 mil |
| Palm Thickness (English) | 7.9 mil |
| Cleanroom Class | Class 10 (ISO 4) |
| Latex Free | No |
| Exterior Finish | Fully Textured |
| Certifications/Compliance | ASTM D3577,ASTM D5151,ASTM D412,ASTM D573,ASTM D3767,ASTM D6319,IEST-RP-CC005,ASTM D5712 |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.20 mm |
Kimtech™ A5 Sterile Cleanroom Sleeve Protector
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Kimtech™ A5 Sterile Cleanroom Sleeve Protector with integrated thumb loop are triple bagged, vacuum packed, and suitable for ISO Class 5 or higher cleanrooms.
Kimtech™ G3 Sterile White Nitrile Gloves
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Nitrile gloves feature a textured finish and provide enhanced sterile cleanroom protection for a wide range of contaminants in a Class 10/ISO 4 critical environment. Hand-specific pairs of Kimtech™ G3 Sterile White Nitrile gloves are appropriate for workers and technicians in medical device manufacturing, pharmaceuticals, or research labs.
| Disposable | Single-use |
|---|---|
| Length (Metric) | 30.5 cm |
| Chemotherapeutic Rated | Yes |
| Finger Thickness (English) | 6 mil |
| Color | White |
| Length (English) | 12 in. |
| Material | Nitrile |
| Powdered | Powder Free |
| Sterility | Sterile |
| Hand | Specific |
| Finger Thickness (Metric) | 0.15 mm |
| Thickness (Metric) | 0.15 mm |
| Accelerator Free | Yes |
| Series | Kimtech™ G3 Sterile |
| Certifications/Compliance (Europe) | EN ISO 374-1:2016; EN 374-4:2003; EN ISO 374-5:2016 |
| Product Type | Cleanroom Gloves |
| Thickness (English) | 6 mil |
| Palm Thickness (English) | 5.1 mil |
| Cleanroom Class | Class 10 (ISO 4) |
| Latex Free | Yes |
| Exterior Finish | Fully Textured |
| Certifications/Compliance | ASTM D5151,ASTM D412,ASTM D573,ASTM D3767,ASTM D3577,ASTM D3767,ASTM D6319,IEST-RP-CC005,ASTM D6978-05 |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.13 mm |
Contec™ PROSAT™ Presaturated Knitted Polynit Wipes
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Clean surfaces and equipment in critical environments where aggressive solvents are needed. The PROSAT™ Presaturated Knitted Polynit Wipes are pre-moistened polyester wipes for general purpose cleanings.
| Disposable | Single-use |
|---|---|
| Construction | No-run pique knit |
| Basis Weight | 140 gsm |
| Product Type | Pre-saturated Wipe |
| Material | Polyester |
| Ply | 1-ply |
| Autoclavable | Not Autoclavable |
Contec™ EasyReach™ Cleaning System: Cleaning Tool
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Passes easily through transfer hatches, requires no assembly and is maintenance-free. EasyReach™ Cleaning System is ideal for isolators, cabinets, laminar air-flow hoods, glove boxes and other mini environments.
| Disposable | Reusable |
|---|---|
| Sterility | Non-sterile |
Ansell BioClean™ Clearview™ Sterile Single Use Cleanroom Goggles
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
BioClean™ Clearview™ Sterile Single Use Cleanroom Goggles for safety in cleanroom environments are anti-fogging and optically correct for clear vision.
Contec™ Sterile 70% Isopropanol
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Contec™ Sterile 70% Isopropanol is ideal for wiping down of isolators, cabinets, laminar flow hoods and surfaces.
| Composition | 70% USP Isopropyl Alcohol and 30% USP Purified Water |
|---|---|
| Form | Liquid |
| Certifications/Compliance | USP 797 |
| For Use With (Application) | Wipe down of isolators, cabinets, laminar flow hoods and surfaces |
| Appearance | Clear |
| Sterility | Sterile |
Berkshire™ 360 Isolator Cleaning System
Quick, easy and efficient cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
| For Use With (Application) | Cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments. |
|---|
VAI DECON-QUAT 200C Fifth Generation Quaternary Ammonium Solution, Sterile
DECON-QUAT 200C Fifth Generation Quaternary Ammonium Solution is formulated to disinfect, clean, and sanitize hard, non-porous surfaces.
| Product Type | Quaternary Ammonium |
|---|---|
| Sterility | Sterile |
Micronova™ ACME Thread Plastic Handle
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Plastic coated fiberglass, fixed length and extendable handles. Designed with ACME Connect threaded ends for use with the entire range of Micronova adapters, mop heads and tools for simplified and versatile cleaning.
| Disposable | Reusable |
|---|---|
| Care Instructions | Store at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage |
| For Use With (Equipment) | QP mop adapters and brushes with threaded connection |
| Color-coded | Yes |
| Quantity | 1 Ea. |
| Material | Fiberglass,Plastic |
| Resistant To | Common disinfectants and cleaning solutions |
| Certifications/Compliance | ISO 9001 Certified Facility Certificate of Conformance |
| For Use With (Application) | Mop Handle for Cleaning and Sanitizing Cleanrooms |
| Autoclavable | Autoclavable |
| Sterility | Non-sterile |
Hourglass International™ HandPRO™ 617200 Sterile Nitrile 16 in. Clean Class Gloves
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Sterile nitrile gloves are thermoform packed, CE triple-bagged, and gamma sterilized for compatibility with controlled environments ISO 5 and higher.
| Disposable | Single-use |
|---|---|
| Chlorinated | No |
| Length (Metric) | 40 cm |
| Packaging Type | Pair Packed, Triple Bagged |
| Finger Thickness (English) | 4.7 to 5.5 mil |
| Color | White |
| Length (English) | 16 in. |
| Material | Nitrile |
| Powdered | Powder Free |
| Sterility | Sterile |
| Hand | Ambidextrous |
| Finger Thickness (Metric) | 0.12 to 0.14 mm |
| Thickness (Metric) | 0.12 to 0.14 mm |
| Accelerator Free | No |
| Product Type | Controlled Environment Gloves |
| Thickness (English) | 4.7 to 5.5 mil |
| Palm Thickness (English) | 3.9 to 4.7 mil |
| Cleanroom Class | Class 100 (ISO 5) |
| Latex Free | Yes |
| Exterior Finish | Smooth |
| Manufacturing/QMS Audit Standards | ISO 13485, ISO 9001 |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.10 to 0.12 mm |
Micronova™ Isolator Tool and Covers
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
The angled head adapter with a 360° swivel works with a range of mop covers to efficiently clean and disinfect isolators, hoods, RABS and small equipment in clean and controlled environments.
| Care Instructions | Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C. |
|---|---|
| Width (English) | 2 in. |
| Length (Metric) | 22.9 cm |
| Product Type | Isolator Cleaning Tool Cover |
| Color-coded | No |
| Length (English) | 9 in. |
| Dimensions | 22.9 x 5.1 cm (9 x 2 in.) |
| Resistant To | Chemicals |
| Autoclavable | Autoclavable |
| Width (Metric) | 5.1 cm |
| Sterility | Non-sterile |
Ansell BioClean™ N-Plus BNPS 16 in. Sterile Nitrile Cleanroom Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Elbow-length nitrile gloves offer resistance to a range of chemicals and are hand specific to enable prolonged use. Recommended for Class 10/ISO 4 critical environments.
| Disposable | Single-use |
|---|---|
| Length (Metric) | 40 cm |
| Finger Thickness (English) | 7.8 mil |
| Color | White |
| Length (English) | 16 in. |
| Material | Nitrile |
| Powdered | Powder Free |
| Sterility | Sterile |
| Hand | Specific |
| Finger Thickness (Metric) | 0.20 mm |
| Thickness (Metric) | 0.20 mm |
| Series | BioClean™ N-Plus BNPS |
| Certifications/Compliance (Europe) | CE 0598, EN 420:2003 + A1:2009, EN 421:2010, EN 455 Part 2, EN ISO 374-1:2016, EN ISO 374-5:2016, Food Contact, Category III |
| Thickness (English) | 7.8 mil |
| Palm Thickness (English) | 6.6 mil |
| Cleanroom Class | Class 10 (ISO 4) |
| Latex Free | Yes |
| Exterior Finish | Fully Textured |
| Certifications/Compliance | ASTM D6978-05 |
| Manufacturing/QMS Audit Standards | ISO 9001, PPE Regulation 2016 425 Module D |
| AQL | 0.65 |
| Palm Thickness (Metric) | 0.17 mm |
VAI DECON-CLEAN Residue Cleaner
DECON-CLEAN Residue Cleaner is designed to remove residues from disinfecting agents.
| Includes | Certificate of analysis, certificate of irradiation and certificate of sterility |
|---|---|
| Filtered | Filtered |
| Certifications/Compliance | USP, ISO |
| For Use With (Application) | Removes residues from sanitizers, disinfectants and sporicides, used for walls, ceilings, floors, stainless items, countertops, aseptic connections etc. |
VAI STERI-PEROX Saturated Wipes
STERI-PEROX Saturated Wipes can be used for both aseptic and non-aseptic wipe downs.