Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAUGUAACAUGGUCCACUAACU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-379-3p | MIMAT0004690 |
| Mouse | mmu-miR-379-3p | MIMAT0017080 |
| Sheep | oar-miR-379-3p | MIMAT0019248 |
Stem-loop Details
Human
Mouse
Sheep
More Information
Related Products
miRBase Alias:
Human: hsa-miR-379* (v17)
Mouse: mmu-miR-379* (v17)
Mouse: mmu-miR-379* (v17)

Genomic Map