Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. 12 - 109709462 - 109709462 [+] on 109709462 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-379-3p | MIMAT0017080 |
| Mouse | mmu-miR-379-5p | MIMAT0000743 |
Stem-loop Details
Mouse

Genomic Map