Assay Details
Amplicon Length: | 61 |
Chromosome Location: | Chr. 14 - 101021867 - 101021867 [-] on 101021867 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-379-3p | MIMAT0004690 |
| Human | hsa-miR-379-5p | MIMAT0000733 |
Stem-loop Details
Human

Genomic Map