Assay Details
Amplicon Length: | 105 |
Chromosome Location: | Chr. X - 59098160 - 59098160 [-] on 59098160 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-504-3p | MIMAT0017277 |
| Mouse | mmu-miR-504-5p | MIMAT0004889 |
Stem-loop Details
Mouse

Genomic Map