Assay Details
Gene Family ID: | MIPF0000437, mir-504 |
Mature miRNA Sequence: | AGACCCUGGUCUGCACUCUAUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-504-5p | MIMAT0002875 |
| Mouse | mmu-miR-504-5p | MIMAT0004889 |
| Dog | cfa-miR-504 | MIMAT0009907 |
| Guinea pig | cpo-miR-504-5p | MIMAT0047309 |
| Horse | eca-miR-504 | MIMAT0013228 |
| Pan troglodytes | ptr-miR-504 | MIMAT0008177 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-504 | MIMAT0015927 |
| Rabbit | ocu-miR-504-5p | MIMAT0048501 |
| Rhesus monkey | mml-miR-504-5p | MIMAT0006368 |
| Tupaia chinensis | tch-miR-504 | MIMAT0036605 |
Stem-loop Details
Human
Mouse
Dog
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
Tupaia chinensis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-504 (v19)
Mouse: mmu-miR-504 (v17)
Macaca mulatta: mml-miR-504 (v19)
Mouse: mmu-miR-504 (v17)
Macaca mulatta: mml-miR-504 (v19)

Genomic Map