Assay Details
Gene Family ID: | MIPF0000437, mir-504 |
Mature miRNA Sequence: | AGACCCUGGUCUGCACUCUAUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-504-5p | MIMAT0002875 | |
| Mouse | mmu-miR-504-5p | MIMAT0004889 | |
| Dog | cfa-miR-504 | MIMAT0009907 | |
| Guinea pig | cpo-miR-504-5p | MIMAT0047309 | |
| Horse | eca-miR-504 | MIMAT0013228 | |
| Pan troglodytes | ptr-miR-504 | MIMAT0008177 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-504 | MIMAT0015927 | |
| Rabbit | ocu-miR-504-5p | MIMAT0048501 | |
| Rhesus monkey | mml-miR-504-5p | MIMAT0006368 | |
| Tupaia chinensis | tch-miR-504 | MIMAT0036605 | |
Stem-loop Details
Human
Mouse
Dog
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
Tupaia chinensis
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-504 (v19)
Mouse: mmu-miR-504 (v17)
Macaca mulatta: mml-miR-504 (v19)
Mouse: mmu-miR-504 (v17)
Macaca mulatta: mml-miR-504 (v19)

Genomic Map