Assay Details
Gene Family ID: | MIPF0000212, mir-540 |
Mature miRNA Sequence: | CAAGGGUCACCCUCUGACUCUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-540-5p | MIMAT0004786 |
| Rat | rno-miR-540-5p | MIMAT0017211 |
Stem-loop Details
Mouse
Rat

Genomic Map