Assay Details
Mature miRNA Sequence: | UACCCUGUAGAACCGAAUUUGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Alligator mississippiensis | ami-miR-10b-5p | MIMAT0038032 | |
Anolis carolinensis | aca-miR-10b-5p | MIMAT0021719 | |
Astatotilapia burtoni | abu-miR-10a | MIMAT0042039 | |
Callithrix jacchus | cja-miR-10b | MIMAT0039433 | |
Chicken | gga-miR-10b-5p | MIMAT0001148 | |
Chrysemys picta | cpi-miR-10b-5p | MIMAT0037627 | |
View More Rows | |||
Columba livia | cli-miR-10b-5p | MIMAT0038406 | |
Cricetulus griseus | cgr-miR-10b-5p | MIMAT0023737 | |
Cyprinus carpio | ccr-miR-10b | MIMAT0026200 | |
Daubentonia madagascariensis | dma-miR-10 | MIMAT0049297 | |
Gadus morhua | gmo-miR-10a-5p | MIMAT0044088 | |
Goat | chi-miR-10b-5p | MIMAT0035913 | |
Gorilla gorilla | ggo-miR-10b | MIMAT0002491 | |
Ictalurus punctatus | ipu-miR-10b | MIMAT0029392 | |
Macaca nemestrina | mne-miR-10b | MIMAT0002492 | |
Metriaclima zebra | mze-miR-10a | MIMAT0042276 | |
Microcebus murinus | mmr-miR-10 | MIMAT0049206 | |
Neolamprologus brichardi | nbr-miR-10a | MIMAT0042465 | |
Nomascus leucogenys | nle-miR-10b | MIMAT0049465 | |
Oreochromis niloticus | oni-miR-10a | MIMAT0042642 | |
Ornithorhynchus anatinus | oan-miR-10b-5p | MIMAT0007218 | |
Oryzias latipes | ola-miR-10b | MIMAT0022651 | |
Otolemur garnettii | oga-miR-10 | MIMAT0049631 | |
Pan paniscus | ppa-miR-10b | MIMAT0002493 | |
Papio hamadryas | pha-miR-10b | MIMAT0049592 | |
Pig | ssc-miR-10b | MIMAT0013885 | |
Pteropus alecto | pal-miR-10b-5p | MIMAT0039899 | |
Pundamilia nyererei | pny-miR-10a | MIMAT0042852 | |
Python bivittatus | pbv-miR-10b-5p | MIMAT0038826 | |
Saimiri boliviensis | sbo-miR-10b | MIMAT0049503 | |
Salmo salar | ssa-miR-10b-5p | MIMAT0032295 | |
Taeniopygia guttata | tgu-miR-10a-5p | MIMAT0018618 | |
Xenopus laevis | xla-miR-10b-5p | MIMAT0046424 | |
Xenopus tropicalis | xtr-miR-10b | MIMAT0003558 |
Stem-loop Details
Alligator mississippiensis
Stem-loop ID | ami-mir-10b |
Stem-loop Accession # | MI0029664 |
Stem-loop Sequence | CGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038032
miRBase ID: ami-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | aca-mir-10b |
Stem-loop Accession # | MI0018717 |
Stem-loop Sequence | GUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUUACACAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0021719
miRBase ID: aca-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | abu-mir-10a-1 |
Stem-loop Accession # | MI0033895 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042039
miRBase ID: abu-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | abu-mir-10a-2 |
Stem-loop Accession # | MI0033713 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042039
miRBase ID: abu-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | cja-mir-10b |
Stem-loop Accession # | MI0031954 |
Stem-loop Sequence | GGGACCCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCACGUUUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0039433
miRBase ID: cja-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | gga-mir-10b |
Stem-loop Accession # | MI0001216 |
Stem-loop Sequence | CAGAACGUUAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAUAUUCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0001148
miRBase ID: gga-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | cpi-mir-10b |
Stem-loop Accession # | MI0029396 |
Stem-loop Sequence | UAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0037627
miRBase ID: cpi-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | cli-mir-10b |
Stem-loop Accession # | MI0029904 |
Stem-loop Sequence | UUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAUAUUCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGAAA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038406
miRBase ID: cli-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | cgr-mir-10b |
Stem-loop Accession # | MI0020382 |
Stem-loop Sequence | UGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUACCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0023737
miRBase ID: cgr-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ccr-mir-10b |
Stem-loop Accession # | MI0023308 |
Stem-loop Sequence | AAAGGCCUGUCAGUAGUCGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUCCAAAACAUGAAAAUCGCAAAUACGUCUCUACAGGAAUACAUGGGCGACGUAAAAGCCACG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0026200
miRBase ID: ccr-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | dma-mir-10 |
Stem-loop Accession # | MI0040032 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049297
miRBase ID: dma-miR-10 Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | gmo-mir-10a-1 |
Stem-loop Accession # | MI0036031 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAGUCACAGAUUCGAUUCUAGGGGAGUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0044088
miRBase ID: gmo-miR-10a-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | gmo-mir-10a-2 |
Stem-loop Accession # | MI0036078 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUUCAGUACAUAUCAUCGCAAAUACGUCUCUACAGGAA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0044088
miRBase ID: gmo-miR-10a-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | chi-mir-10b |
Stem-loop Accession # | MI0030597 |
Stem-loop Sequence | CAGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCUCGUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCAC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0035913
miRBase ID: chi-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ggo-mir-10b |
Stem-loop Accession # | MI0002793 |
Stem-loop Sequence | CCAGACAUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002491
miRBase ID: ggo-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ipu-mir-10b-1 |
Stem-loop Accession # | MI0024465 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAAAAAUAACAUUCACAGAUUCGAUUCUAGGGGAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0029392
miRBase ID: ipu-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ipu-mir-10b-2 |
Stem-loop Accession # | MI0024466 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUCCAAAACAUCAAAAUCGCAAAUACGUCUCUACAGGAAUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0029392
miRBase ID: ipu-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | mne-mir-10b |
Stem-loop Accession # | MI0002794 |
Stem-loop Sequence | CAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002492
miRBase ID: mne-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | mze-mir-10a |
Stem-loop Accession # | MI0034011 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042276
miRBase ID: mze-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | mmr-mir-10 |
Stem-loop Accession # | MI0039928 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACGUAGUCACAGAUUCGAUUCUAGGGGAAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049206
miRBase ID: mmr-miR-10 Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | nbr-mir-10a-1 |
Stem-loop Accession # | MI0034272 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUUUAGGGGAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042465
miRBase ID: nbr-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | nbr-mir-10a-2 |
Stem-loop Accession # | MI0034273 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042465
miRBase ID: nbr-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | nle-mir-10b |
Stem-loop Accession # | MI0040219 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049465
miRBase ID: nle-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | oni-mir-10a-1 |
Stem-loop Accession # | MI0034518 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042642
miRBase ID: oni-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | oni-mir-10a-2 |
Stem-loop Accession # | MI0034519 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042642
miRBase ID: oni-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | oan-mir-10b |
Stem-loop Accession # | MI0006921 |
Stem-loop Sequence | GAAUAUUAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0007218
miRBase ID: oan-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ola-mir-10b |
Stem-loop Accession # | MI0019535 |
Stem-loop Sequence | UCGUGGCUUCCCGUCGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAAGAACGCUCAAUCACAGAUUCGAUUCUAGGGGAGUAUAUGGUCGAUGAAAAAACUUCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0022651
miRBase ID: ola-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | oga-mir-10 |
Stem-loop Accession # | MI0040414 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049631
miRBase ID: oga-miR-10 Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ppa-mir-10b |
Stem-loop Accession # | MI0002795 |
Stem-loop Sequence | CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCGUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002493
miRBase ID: ppa-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | pha-mir-10b |
Stem-loop Accession # | MI0040371 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049592
miRBase ID: pha-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ssc-mir-10b |
Stem-loop Accession # | MI0013102 |
Stem-loop Sequence | GUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0013885
miRBase ID: ssc-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | pal-mir-10b |
Stem-loop Accession # | MI0032390 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0039899
miRBase ID: pal-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | pny-mir-10a-1 |
Stem-loop Accession # | MI0034790 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042852
miRBase ID: pny-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | pny-mir-10a-2 |
Stem-loop Accession # | MI0034791 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042852
miRBase ID: pny-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | pbv-mir-10b-1 |
Stem-loop Accession # | MI0030154 |
Stem-loop Sequence | ACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUUACGUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038826
miRBase ID: pbv-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | pbv-mir-10b-2 |
Stem-loop Accession # | MI0030155 |
Stem-loop Sequence | CAUCGUUUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAGCACCUCUAUUGCAAAUUCGUCUCUAGGGGGAUAUGUGGAUGAUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038826
miRBase ID: pbv-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | sbo-mir-10b |
Stem-loop Accession # | MI0040268 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049503
miRBase ID: sbo-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ssa-mir-10b-1 |
Stem-loop Accession # | MI0026444 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUUAAUAGAUAACAAUCGCAAGUACGUCUCUACAGGAAUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032295
miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ssa-mir-10b-2 |
Stem-loop Accession # | MI0026445 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUAAAACAAGUCACAGAUUCGAUUCUAGGGGAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032295
miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ssa-mir-10b-3 |
Stem-loop Accession # | MI0026446 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUAAAACAACAGUCACAGAUUCGAUUCUAGGGGAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032295
miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | ssa-mir-10b-4 |
Stem-loop Accession # | MI0026447 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUUAAUAUAUUACAAUCGCAAGUACGUCUCUACAGGAAUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032295
miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | tgu-mir-10a |
Stem-loop Accession # | MI0016248 |
Stem-loop Sequence | ACAGAAUGUUAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAUAUUCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCAC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0018618
miRBase ID: tgu-miR-10a-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | xla-mir-10b |
Stem-loop Accession # | MI0038234 |
Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUUUUAACAUAGUCACAGAUUCGAUUCUAGGGGGAUA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0046424
miRBase ID: xla-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
Stem-loop ID | xtr-mir-10b |
Stem-loop Accession # | MI0004797 |
Stem-loop Sequence | AACGUUGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUGGUUCGUACAGUCACAGAUUCGAUUCUAGGGGGAUAUAUGGUCGAUGCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003558
miRBase ID: xtr-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |

More Information
Related Products




miRBase Alias:
Human: hsa-miR-10b (v9.2)
Aeromonas caviae Ae398: aca-miR-10b (v18)
Gallus gallus: gga-miR-10b (v19)
Ochrobactrum anthropi: oan-miR-10b (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-10a (v19)
Aeromonas caviae Ae398: aca-miR-10b (v18)
Gallus gallus: gga-miR-10b (v19)
Ochrobactrum anthropi: oan-miR-10b (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-10a (v19)
