Assay Details
Mature miRNA Sequence: | UACCCUGUAGAACCGAAUUUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-10b-5p | MIMAT0038032 |
| Anolis carolinensis | aca-miR-10b-5p | MIMAT0021719 |
| Astatotilapia burtoni | abu-miR-10a | MIMAT0042039 |
| Callithrix jacchus | cja-miR-10b | MIMAT0039433 |
| Chicken | gga-miR-10b-5p | MIMAT0001148 |
| Chrysemys picta | cpi-miR-10b-5p | MIMAT0037627 |
| View More Rows | ||
| Columba livia | cli-miR-10b-5p | MIMAT0038406 |
| Cricetulus griseus | cgr-miR-10b-5p | MIMAT0023737 |
| Cyprinus carpio | ccr-miR-10b | MIMAT0026200 |
| Daubentonia madagascariensis | dma-miR-10 | MIMAT0049297 |
| Gadus morhua | gmo-miR-10a-5p | MIMAT0044088 |
| Goat | chi-miR-10b-5p | MIMAT0035913 |
| Gorilla gorilla | ggo-miR-10b | MIMAT0002491 |
| Ictalurus punctatus | ipu-miR-10b | MIMAT0029392 |
| Macaca nemestrina | mne-miR-10b | MIMAT0002492 |
| Metriaclima zebra | mze-miR-10a | MIMAT0042276 |
| Microcebus murinus | mmr-miR-10 | MIMAT0049206 |
| Neolamprologus brichardi | nbr-miR-10a | MIMAT0042465 |
| Nomascus leucogenys | nle-miR-10b | MIMAT0049465 |
| Oreochromis niloticus | oni-miR-10a | MIMAT0042642 |
| Ornithorhynchus anatinus | oan-miR-10b-5p | MIMAT0007218 |
| Oryzias latipes | ola-miR-10b | MIMAT0022651 |
| Otolemur garnettii | oga-miR-10 | MIMAT0049631 |
| Pan paniscus | ppa-miR-10b | MIMAT0002493 |
| Papio hamadryas | pha-miR-10b | MIMAT0049592 |
| Pig | ssc-miR-10b | MIMAT0013885 |
| Pteropus alecto | pal-miR-10b-5p | MIMAT0039899 |
| Pundamilia nyererei | pny-miR-10a | MIMAT0042852 |
| Python bivittatus | pbv-miR-10b-5p | MIMAT0038826 |
| Saimiri boliviensis | sbo-miR-10b | MIMAT0049503 |
| Salmo salar | ssa-miR-10b-5p | MIMAT0032295 |
| Taeniopygia guttata | tgu-miR-10a-5p | MIMAT0018618 |
| Xenopus laevis | xla-miR-10b-5p | MIMAT0046424 |
| Xenopus tropicalis | xtr-miR-10b | MIMAT0003558 |
Stem-loop Details
Alligator mississippiensis
| Stem-loop ID | ami-mir-10b |
| Stem-loop Accession # | MI0029664 |
| Stem-loop Sequence | CGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038032
miRBase ID: ami-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | aca-mir-10b |
| Stem-loop Accession # | MI0018717 |
| Stem-loop Sequence | GUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUUACACAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0021719
miRBase ID: aca-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | abu-mir-10a-1 |
| Stem-loop Accession # | MI0033895 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042039
miRBase ID: abu-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | abu-mir-10a-2 |
| Stem-loop Accession # | MI0033713 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042039
miRBase ID: abu-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | cja-mir-10b |
| Stem-loop Accession # | MI0031954 |
| Stem-loop Sequence | GGGACCCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCACGUUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039433
miRBase ID: cja-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | gga-mir-10b |
| Stem-loop Accession # | MI0001216 |
| Stem-loop Sequence | CAGAACGUUAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAUAUUCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001148
miRBase ID: gga-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | cpi-mir-10b |
| Stem-loop Accession # | MI0029396 |
| Stem-loop Sequence | UAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037627
miRBase ID: cpi-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | cli-mir-10b |
| Stem-loop Accession # | MI0029904 |
| Stem-loop Sequence | UUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAUAUUCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038406
miRBase ID: cli-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | cgr-mir-10b |
| Stem-loop Accession # | MI0020382 |
| Stem-loop Sequence | UGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUACCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0023737
miRBase ID: cgr-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ccr-mir-10b |
| Stem-loop Accession # | MI0023308 |
| Stem-loop Sequence | AAAGGCCUGUCAGUAGUCGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUCCAAAACAUGAAAAUCGCAAAUACGUCUCUACAGGAAUACAUGGGCGACGUAAAAGCCACG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0026200
miRBase ID: ccr-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | dma-mir-10 |
| Stem-loop Accession # | MI0040032 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049297
miRBase ID: dma-miR-10 Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-10a-1 |
| Stem-loop Accession # | MI0036031 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAGUCACAGAUUCGAUUCUAGGGGAGUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044088
miRBase ID: gmo-miR-10a-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-10a-2 |
| Stem-loop Accession # | MI0036078 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUUCAGUACAUAUCAUCGCAAAUACGUCUCUACAGGAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044088
miRBase ID: gmo-miR-10a-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | chi-mir-10b |
| Stem-loop Accession # | MI0030597 |
| Stem-loop Sequence | CAGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCUCGUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0035913
miRBase ID: chi-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ggo-mir-10b |
| Stem-loop Accession # | MI0002793 |
| Stem-loop Sequence | CCAGACAUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002491
miRBase ID: ggo-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ipu-mir-10b-1 |
| Stem-loop Accession # | MI0024465 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAAAAAUAACAUUCACAGAUUCGAUUCUAGGGGAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0029392
miRBase ID: ipu-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ipu-mir-10b-2 |
| Stem-loop Accession # | MI0024466 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUCCAAAACAUCAAAAUCGCAAAUACGUCUCUACAGGAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0029392
miRBase ID: ipu-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | mne-mir-10b |
| Stem-loop Accession # | MI0002794 |
| Stem-loop Sequence | CAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002492
miRBase ID: mne-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | mze-mir-10a |
| Stem-loop Accession # | MI0034011 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042276
miRBase ID: mze-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | mmr-mir-10 |
| Stem-loop Accession # | MI0039928 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACGUAGUCACAGAUUCGAUUCUAGGGGAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049206
miRBase ID: mmr-miR-10 Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | nbr-mir-10a-1 |
| Stem-loop Accession # | MI0034272 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUUUAGGGGAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042465
miRBase ID: nbr-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | nbr-mir-10a-2 |
| Stem-loop Accession # | MI0034273 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042465
miRBase ID: nbr-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | nle-mir-10b |
| Stem-loop Accession # | MI0040219 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049465
miRBase ID: nle-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | oni-mir-10a-1 |
| Stem-loop Accession # | MI0034518 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042642
miRBase ID: oni-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | oni-mir-10a-2 |
| Stem-loop Accession # | MI0034519 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042642
miRBase ID: oni-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | oan-mir-10b |
| Stem-loop Accession # | MI0006921 |
| Stem-loop Sequence | GAAUAUUAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0007218
miRBase ID: oan-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ola-mir-10b |
| Stem-loop Accession # | MI0019535 |
| Stem-loop Sequence | UCGUGGCUUCCCGUCGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAAGAACGCUCAAUCACAGAUUCGAUUCUAGGGGAGUAUAUGGUCGAUGAAAAAACUUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0022651
miRBase ID: ola-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | oga-mir-10 |
| Stem-loop Accession # | MI0040414 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049631
miRBase ID: oga-miR-10 Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ppa-mir-10b |
| Stem-loop Accession # | MI0002795 |
| Stem-loop Sequence | CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCGUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002493
miRBase ID: ppa-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | pha-mir-10b |
| Stem-loop Accession # | MI0040371 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049592
miRBase ID: pha-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ssc-mir-10b |
| Stem-loop Accession # | MI0013102 |
| Stem-loop Sequence | GUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013885
miRBase ID: ssc-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | pal-mir-10b |
| Stem-loop Accession # | MI0032390 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039899
miRBase ID: pal-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | pny-mir-10a-1 |
| Stem-loop Accession # | MI0034790 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042852
miRBase ID: pny-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | pny-mir-10a-2 |
| Stem-loop Accession # | MI0034791 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042852
miRBase ID: pny-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | pbv-mir-10b-1 |
| Stem-loop Accession # | MI0030154 |
| Stem-loop Sequence | ACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUUACGUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038826
miRBase ID: pbv-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | pbv-mir-10b-2 |
| Stem-loop Accession # | MI0030155 |
| Stem-loop Sequence | CAUCGUUUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAGCACCUCUAUUGCAAAUUCGUCUCUAGGGGGAUAUGUGGAUGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038826
miRBase ID: pbv-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | sbo-mir-10b |
| Stem-loop Accession # | MI0040268 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049503
miRBase ID: sbo-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-10b-1 |
| Stem-loop Accession # | MI0026444 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUUAAUAGAUAACAAUCGCAAGUACGUCUCUACAGGAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032295
miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-10b-2 |
| Stem-loop Accession # | MI0026445 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUAAAACAAGUCACAGAUUCGAUUCUAGGGGAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032295
miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-10b-3 |
| Stem-loop Accession # | MI0026446 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUAAAACAACAGUCACAGAUUCGAUUCUAGGGGAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032295
miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-10b-4 |
| Stem-loop Accession # | MI0026447 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUUAAUAUAUUACAAUCGCAAGUACGUCUCUACAGGAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032295
miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | tgu-mir-10a |
| Stem-loop Accession # | MI0016248 |
| Stem-loop Sequence | ACAGAAUGUUAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAUAUUCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0018618
miRBase ID: tgu-miR-10a-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | xla-mir-10b |
| Stem-loop Accession # | MI0038234 |
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUUUUAACAUAGUCACAGAUUCGAUUCUAGGGGGAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046424
miRBase ID: xla-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
| Stem-loop ID | xtr-mir-10b |
| Stem-loop Accession # | MI0004797 |
| Stem-loop Sequence | AACGUUGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUGGUUCGUACAGUCACAGAUUCGAUUCUAGGGGGAUAUAUGGUCGAUGCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003558
miRBase ID: xtr-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA |
More Information
Related Products
miRBase Alias:
Human: hsa-miR-10b (v9.2)
Aeromonas caviae Ae398: aca-miR-10b (v18)
Gallus gallus: gga-miR-10b (v19)
Ochrobactrum anthropi: oan-miR-10b (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-10a (v19)
Aeromonas caviae Ae398: aca-miR-10b (v18)
Gallus gallus: gga-miR-10b (v19)
Ochrobactrum anthropi: oan-miR-10b (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-10a (v19)

Genomic Map