Assay Details
| Mature miRNA Sequence: | UACCCUGUAGAACCGAAUUUGU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Alligator mississippiensis | ami-miR-10b-5p | MIMAT0038032 | 
| Anolis carolinensis | aca-miR-10b-5p | MIMAT0021719 | 
| Astatotilapia burtoni | abu-miR-10a | MIMAT0042039 | 
| Callithrix jacchus | cja-miR-10b | MIMAT0039433 | 
| Chicken | gga-miR-10b-5p | MIMAT0001148 | 
| Chrysemys picta | cpi-miR-10b-5p | MIMAT0037627 | 
| View More Rows | ||
| Columba livia | cli-miR-10b-5p | MIMAT0038406 | 
| Cricetulus griseus | cgr-miR-10b-5p | MIMAT0023737 | 
| Cyprinus carpio | ccr-miR-10b | MIMAT0026200 | 
| Daubentonia madagascariensis | dma-miR-10 | MIMAT0049297 | 
| Gadus morhua | gmo-miR-10a-5p | MIMAT0044088 | 
| Goat | chi-miR-10b-5p | MIMAT0035913 | 
| Gorilla gorilla | ggo-miR-10b | MIMAT0002491 | 
| Ictalurus punctatus | ipu-miR-10b | MIMAT0029392 | 
| Macaca nemestrina | mne-miR-10b | MIMAT0002492 | 
| Metriaclima zebra | mze-miR-10a | MIMAT0042276 | 
| Microcebus murinus | mmr-miR-10 | MIMAT0049206 | 
| Neolamprologus brichardi | nbr-miR-10a | MIMAT0042465 | 
| Nomascus leucogenys | nle-miR-10b | MIMAT0049465 | 
| Oreochromis niloticus | oni-miR-10a | MIMAT0042642 | 
| Ornithorhynchus anatinus | oan-miR-10b-5p | MIMAT0007218 | 
| Oryzias latipes | ola-miR-10b | MIMAT0022651 | 
| Otolemur garnettii | oga-miR-10 | MIMAT0049631 | 
| Pan paniscus | ppa-miR-10b | MIMAT0002493 | 
| Papio hamadryas | pha-miR-10b | MIMAT0049592 | 
| Pig | ssc-miR-10b | MIMAT0013885 | 
| Pteropus alecto | pal-miR-10b-5p | MIMAT0039899 | 
| Pundamilia nyererei | pny-miR-10a | MIMAT0042852 | 
| Python bivittatus | pbv-miR-10b-5p | MIMAT0038826 | 
| Saimiri boliviensis | sbo-miR-10b | MIMAT0049503 | 
| Salmo salar | ssa-miR-10b-5p | MIMAT0032295 | 
| Taeniopygia guttata | tgu-miR-10a-5p | MIMAT0018618 | 
| Xenopus laevis | xla-miR-10b-5p | MIMAT0046424 | 
| Xenopus tropicalis | xtr-miR-10b | MIMAT0003558 | 
Stem-loop Details
Alligator mississippiensis
| Stem-loop ID | ami-mir-10b | 
| Stem-loop Accession # | MI0029664 | 
| Stem-loop Sequence | CGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0038032 miRBase ID: ami-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | aca-mir-10b | 
| Stem-loop Accession # | MI0018717 | 
| Stem-loop Sequence | GUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUUACACAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0021719 miRBase ID: aca-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | abu-mir-10a-1 | 
| Stem-loop Accession # | MI0033895 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042039 miRBase ID: abu-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | abu-mir-10a-2 | 
| Stem-loop Accession # | MI0033713 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042039 miRBase ID: abu-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | cja-mir-10b | 
| Stem-loop Accession # | MI0031954 | 
| Stem-loop Sequence | GGGACCCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCACGUUUC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0039433 miRBase ID: cja-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | gga-mir-10b | 
| Stem-loop Accession # | MI0001216 | 
| Stem-loop Sequence | CAGAACGUUAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAUAUUCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0001148 miRBase ID: gga-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | cpi-mir-10b | 
| Stem-loop Accession # | MI0029396 | 
| Stem-loop Sequence | UAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0037627 miRBase ID: cpi-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | cli-mir-10b | 
| Stem-loop Accession # | MI0029904 | 
| Stem-loop Sequence | UUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAUAUUCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0038406 miRBase ID: cli-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | cgr-mir-10b | 
| Stem-loop Accession # | MI0020382 | 
| Stem-loop Sequence | UGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUACCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0023737 miRBase ID: cgr-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ccr-mir-10b | 
| Stem-loop Accession # | MI0023308 | 
| Stem-loop Sequence | AAAGGCCUGUCAGUAGUCGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUCCAAAACAUGAAAAUCGCAAAUACGUCUCUACAGGAAUACAUGGGCGACGUAAAAGCCACG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0026200 miRBase ID: ccr-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | dma-mir-10 | 
| Stem-loop Accession # | MI0040032 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049297 miRBase ID: dma-miR-10 Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | gmo-mir-10a-1 | 
| Stem-loop Accession # | MI0036031 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAGUCACAGAUUCGAUUCUAGGGGAGUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0044088 miRBase ID: gmo-miR-10a-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | gmo-mir-10a-2 | 
| Stem-loop Accession # | MI0036078 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUUCAGUACAUAUCAUCGCAAAUACGUCUCUACAGGAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0044088 miRBase ID: gmo-miR-10a-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | chi-mir-10b | 
| Stem-loop Accession # | MI0030597 | 
| Stem-loop Sequence | CAGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCUCGUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCAC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0035913 miRBase ID: chi-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ggo-mir-10b | 
| Stem-loop Accession # | MI0002793 | 
| Stem-loop Sequence | CCAGACAUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0002491 miRBase ID: ggo-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ipu-mir-10b-1 | 
| Stem-loop Accession # | MI0024465 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAAAAAUAACAUUCACAGAUUCGAUUCUAGGGGAGU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0029392 miRBase ID: ipu-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ipu-mir-10b-2 | 
| Stem-loop Accession # | MI0024466 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUCCAAAACAUCAAAAUCGCAAAUACGUCUCUACAGGAAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0029392 miRBase ID: ipu-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | mne-mir-10b | 
| Stem-loop Accession # | MI0002794 | 
| Stem-loop Sequence | CAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0002492 miRBase ID: mne-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | mze-mir-10a | 
| Stem-loop Accession # | MI0034011 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042276 miRBase ID: mze-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | mmr-mir-10 | 
| Stem-loop Accession # | MI0039928 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACGUAGUCACAGAUUCGAUUCUAGGGGAAU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049206 miRBase ID: mmr-miR-10 Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | nbr-mir-10a-1 | 
| Stem-loop Accession # | MI0034272 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUUUAGGGGAGU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042465 miRBase ID: nbr-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | nbr-mir-10a-2 | 
| Stem-loop Accession # | MI0034273 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042465 miRBase ID: nbr-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | nle-mir-10b | 
| Stem-loop Accession # | MI0040219 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049465 miRBase ID: nle-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | oni-mir-10a-1 | 
| Stem-loop Accession # | MI0034518 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042642 miRBase ID: oni-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | oni-mir-10a-2 | 
| Stem-loop Accession # | MI0034519 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042642 miRBase ID: oni-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | oan-mir-10b | 
| Stem-loop Accession # | MI0006921 | 
| Stem-loop Sequence | GAAUAUUAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0007218 miRBase ID: oan-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ola-mir-10b | 
| Stem-loop Accession # | MI0019535 | 
| Stem-loop Sequence | UCGUGGCUUCCCGUCGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAAGAACGCUCAAUCACAGAUUCGAUUCUAGGGGAGUAUAUGGUCGAUGAAAAAACUUCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0022651 miRBase ID: ola-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | oga-mir-10 | 
| Stem-loop Accession # | MI0040414 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049631 miRBase ID: oga-miR-10 Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ppa-mir-10b | 
| Stem-loop Accession # | MI0002795 | 
| Stem-loop Sequence | CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCGUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0002493 miRBase ID: ppa-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | pha-mir-10b | 
| Stem-loop Accession # | MI0040371 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049592 miRBase ID: pha-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ssc-mir-10b | 
| Stem-loop Accession # | MI0013102 | 
| Stem-loop Sequence | GUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0013885 miRBase ID: ssc-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | pal-mir-10b | 
| Stem-loop Accession # | MI0032390 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0039899 miRBase ID: pal-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | pny-mir-10a-1 | 
| Stem-loop Accession # | MI0034790 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUGAAACCAGAUUCACAGAUUCGAUUCUAGGGGAGU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042852 miRBase ID: pny-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | pny-mir-10a-2 | 
| Stem-loop Accession # | MI0034791 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAAGUACAAGCAGUCGCAAAUACGUCUCUACAGGAAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042852 miRBase ID: pny-miR-10a Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | pbv-mir-10b-1 | 
| Stem-loop Accession # | MI0030154 | 
| Stem-loop Sequence | ACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUUACGUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0038826 miRBase ID: pbv-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | pbv-mir-10b-2 | 
| Stem-loop Accession # | MI0030155 | 
| Stem-loop Sequence | CAUCGUUUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAGCACCUCUAUUGCAAAUUCGUCUCUAGGGGGAUAUGUGGAUGAUG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0038826 miRBase ID: pbv-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | sbo-mir-10b | 
| Stem-loop Accession # | MI0040268 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049503 miRBase ID: sbo-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ssa-mir-10b-1 | 
| Stem-loop Accession # | MI0026444 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUUAAUAGAUAACAAUCGCAAGUACGUCUCUACAGGAAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0032295 miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ssa-mir-10b-2 | 
| Stem-loop Accession # | MI0026445 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUAAAACAAGUCACAGAUUCGAUUCUAGGGGAGU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0032295 miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ssa-mir-10b-3 | 
| Stem-loop Accession # | MI0026446 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGAUAAAACAACAGUCACAGAUUCGAUUCUAGGGGAGU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0032295 miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | ssa-mir-10b-4 | 
| Stem-loop Accession # | MI0026447 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUUAAUAUAUUACAAUCGCAAGUACGUCUCUACAGGAAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0032295 miRBase ID: ssa-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | tgu-mir-10a | 
| Stem-loop Accession # | MI0016248 | 
| Stem-loop Sequence | ACAGAAUGUUAUUACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAUAUUCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCAC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0018618 miRBase ID: tgu-miR-10a-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | xla-mir-10b | 
| Stem-loop Accession # | MI0038234 | 
| Stem-loop Sequence | UACCCUGUAGAACCGAAUUUGUGUGGUUUUAACAUAGUCACAGAUUCGAUUCUAGGGGGAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0046424 miRBase ID: xla-miR-10b-5p Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
| Stem-loop ID | xtr-mir-10b | 
| Stem-loop Accession # | MI0004797 | 
| Stem-loop Sequence | AACGUUGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUGGUUCGUACAGUCACAGAUUCGAUUCUAGGGGGAUAUAUGGUCGAUGCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0003558 miRBase ID: xtr-miR-10b Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGU Chromosome Location: NA | 
 Back To Top
 Back To TopMore Information
Related Products
 Ambion® Anti-miR™ miRNA Inhibitor : AM10133
						Ambion® Anti-miR™ miRNA Inhibitor : AM10133 Ambion® Pre-miR™ miRNA Precursor : PM10133
						Ambion® Pre-miR™ miRNA Precursor : PM10133 TaqMan™ MicroRNA Assay : 000388
						TaqMan™ MicroRNA Assay : 000388 mirVana® miRNA inhibitor : MH10133
						mirVana® miRNA inhibitor : MH10133miRBase Alias:
						
							Aeromonas caviae Ae398: aca-miR-10b (v18)
Gallus gallus: gga-miR-10b (v19)
Ochrobactrum anthropi: oan-miR-10b (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-10a (v19)
						
					
				Gallus gallus: gga-miR-10b (v19)
Ochrobactrum anthropi: oan-miR-10b (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-10a (v19)
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top