Assay Details
Gene Family ID: | MIPF0000147, mir-325 |
Mature miRNA Sequence: | CCUAGUAGGUGUCCAGUAAGUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-325 | MIMAT0000771 |
| Pongo pygmaeus | ppy-miR-325 | MIMAT0015829 |
| Rhesus monkey | mml-miR-325 | MIMAT0006268 |
Stem-loop Details
Human
Pongo pygmaeus
Rhesus monkey

Genomic Map