Assay Details
Amplicon Length: | 71 |
Chromosome Location: | Chr. X - 77005105 - 77005105 [-] on 77005105 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-325 | MIMAT0000771 |
Stem-loop Details
Human
