Assay Details
Gene Family ID: | MIPF0000147, mir-325 |
Mature miRNA Sequence: | CCUAGUAGGUGUCCAGUAAGUGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-325 | MIMAT0000771 |
Pongo pygmaeus | ppy-miR-325 | MIMAT0015829 |
Rhesus monkey | mml-miR-325 | MIMAT0006268 |
Stem-loop Details
Human
Pongo pygmaeus
Rhesus monkey
