Assay Details
Gene Family ID: | MIPF0000229, mir-490 |
Mature miRNA Sequence: | CAACCUGGAGGACUCCAUGCUG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-490-3p | MIMAT0002806 | |
Mouse | mmu-miR-490-3p | MIMAT0003780 | |
Rat | rno-miR-490-3p | MIMAT0012823 | |
Alligator mississippiensis | ami-miR-490-3p | MIMAT0038298 | |
Bovine | bta-miR-490 | MIMAT0009331 | |
Chicken | gga-miR-490-3p | MIMAT0003366 | |
View More Rows | |||
Dog | cfa-miR-490 | MIMAT0009875 | |
Gorilla gorilla | ggo-miR-490 | MIMAT0024205 | |
Horse | eca-miR-490-3p | MIMAT0012945 | |
Pan troglodytes | ptr-miR-490 | MIMAT0008165 | |
Pig | ssc-miR-490-3p | MIMAT0022960 | |
Pig | ssc-miR-490 | MIMAT0025376 | |
Pongo pygmaeus | ppy-miR-490-3p | MIMAT0015910 | |
Rhesus monkey | mml-miR-490-3p | MIMAT0006351 |
Stem-loop Details
Human
Mouse
Rat
Alligator mississippiensis
Bovine
Chicken
Dog
Gorilla gorilla
Horse
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-490 (v9.2)
Mouse: mmu-miR-490 (v15)
Rat: rno-miR-490 (v18)
Gallus gallus: gga-miR-490 (v19)
Mouse: mmu-miR-490 (v15)
Rat: rno-miR-490 (v18)
Gallus gallus: gga-miR-490 (v19)
