Assay Details
Gene Family ID: | MIPF0000229, mir-490 | 
Mature miRNA Sequence: | CAACCUGGAGGACUCCAUGCUG | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-490-3p | MIMAT0002806 | 
| Mouse | mmu-miR-490-3p | MIMAT0003780 | 
| Rat | rno-miR-490-3p | MIMAT0012823 | 
| Alligator mississippiensis | ami-miR-490-3p | MIMAT0038298 | 
| Bovine | bta-miR-490 | MIMAT0009331 | 
| Chicken | gga-miR-490-3p | MIMAT0003366 | 
| View More Rows | ||
| Dog | cfa-miR-490 | MIMAT0009875 | 
| Gorilla gorilla | ggo-miR-490 | MIMAT0024205 | 
| Horse | eca-miR-490-3p | MIMAT0012945 | 
| Pan troglodytes | ptr-miR-490 | MIMAT0008165 | 
| Pig | ssc-miR-490-3p | MIMAT0022960 | 
| Pig | ssc-miR-490 | MIMAT0025376 | 
| Pongo pygmaeus | ppy-miR-490-3p | MIMAT0015910 | 
| Rhesus monkey | mml-miR-490-3p | MIMAT0006351 | 
Stem-loop Details
Human
Mouse
Rat
Alligator mississippiensis
Bovine
Chicken
Dog
Gorilla gorilla
Horse
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
						
							Rat: rno-miR-490 (v18)
Gallus gallus: gga-miR-490 (v19)
						
					
				Gallus gallus: gga-miR-490 (v19)

Genomic Map