Assay Details
Gene Family ID: | MIPF0000229, mir-490 |
Mature miRNA Sequence: | CAACCUGGAGGACUCCAUGCUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-490-3p | MIMAT0002806 |
Mouse | mmu-miR-490-3p | MIMAT0003780 |
Rat | rno-miR-490-3p | MIMAT0012823 |
Alligator mississippiensis | ami-miR-490-3p | MIMAT0038298 |
Bovine | bta-miR-490 | MIMAT0009331 |
Chicken | gga-miR-490-3p | MIMAT0003366 |
View More Rows | ||
Dog | cfa-miR-490 | MIMAT0009875 |
Gorilla gorilla | ggo-miR-490 | MIMAT0024205 |
Horse | eca-miR-490-3p | MIMAT0012945 |
Pan troglodytes | ptr-miR-490 | MIMAT0008165 |
Pig | ssc-miR-490-3p | MIMAT0022960 |
Pig | ssc-miR-490 | MIMAT0025376 |
Pongo pygmaeus | ppy-miR-490-3p | MIMAT0015910 |
Rhesus monkey | mml-miR-490-3p | MIMAT0006351 |
Stem-loop Details
Human
Mouse
Rat
Alligator mississippiensis
Bovine
Chicken
Dog
Gorilla gorilla
Horse
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey
