Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CUCCAGAGGGAAGUACUUUCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-520a-5p | MIMAT0002833 |
| Pongo pygmaeus | ppy-miR-520a-5p | MIMAT0015966 |
Stem-loop Details
Human
Pongo pygmaeus

Genomic Map