Assay Details
Amplicon Length: | 113 |
Chromosome Location: | Chr. 19 - 53691137 - 53691137 [-] on 53691137 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-520a-5p | MIMAT0002833 |
Human | hsa-miR-520a-3p | MIMAT0002834 |
Stem-loop Details
Human
