Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CUCCAGAGGGAAGUACUUUCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-520a-5p | MIMAT0002833 |
Pongo pygmaeus | ppy-miR-520a-5p | MIMAT0015966 |
Stem-loop Details
Human
Pongo pygmaeus
