Assay Details
Gene Family ID: | MIPF0000508, mir-631 |
Mature miRNA Sequence: | AGACCUGGCCCAGACCUCAGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-631 | MIMAT0003300 |
| Callithrix jacchus | cja-miR-631 | MIMAT0039591 |
| Pongo pygmaeus | ppy-miR-631 | MIMAT0016070 |
| Rhesus monkey | mml-miR-631 | MIMAT0006476 |
Stem-loop Details
Human
Callithrix jacchus
Pongo pygmaeus
Rhesus monkey

Genomic Map