Assay Details
Gene Family ID: | MIPF0000508, mir-631 |
Mature miRNA Sequence: | AGACCUGGCCCAGACCUCAGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-631 | MIMAT0003300 |
Callithrix jacchus | cja-miR-631 | MIMAT0039591 |
Pongo pygmaeus | ppy-miR-631 | MIMAT0016070 |
Rhesus monkey | mml-miR-631 | MIMAT0006476 |
Stem-loop Details
Human
Callithrix jacchus
Pongo pygmaeus
Rhesus monkey
